Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018845.hg.1 |
TC09001507.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018845.hg.1 |
ENST00000259410.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018845.hg.1 |
ENST00000374118.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018845.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018845.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000473174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000473174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000473174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000473174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000473174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000473174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000473174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000473174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000473174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000491059.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000491059.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000491059.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000491059.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000498016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000498016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000498016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000498016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000498016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
ENST00000498016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900176883.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176883.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176883.hg.1 |
ENST00000473174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176883.hg.1 |
ENST00000491059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176883.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900176883.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176883.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176883.hg.1 |
ENST00000473174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176883.hg.1 |
ENST00000491059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176883.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900176883.hg.1 |
TC0900011277.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900176885.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176885.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176885.hg.1 |
ENST00000491059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176885.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900176885.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176885.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176885.hg.1 |
ENST00000491059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176885.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900176885.hg.1 |
TC0900011277.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000374118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000468460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000473174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000473174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000473174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000473174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000473174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000473174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000473174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000473174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000473174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000491059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000491059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000491059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000491059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000498016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000498016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000498016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000498016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000498016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
ENST00000498016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011277.hg.1 |
NM_138424.1 |
Refseq |