Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000517435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000517435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000517435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000522319.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000522319.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000522319.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
ENST00000522319.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08008364.hg.1 |
TC08000602.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08008364.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08008364.hg.1 |
ENST00000401707.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08008364.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08008364.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08008364.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800157397.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800157397.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800157397.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800157397.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800157397.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800157397.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800157397.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800157397.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800157397.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800157397.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800157397.hg.1 |
TC0800008330.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000349693.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000401707.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000517435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000517435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000517435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000522319.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000522319.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000522319.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
ENST00000522319.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145860.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_001145861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008330.hg.1 |
NM_015029.2 |
Refseq |