Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04020936.hg.1 |
TC04001476.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04020936.hg.1 |
ENST00000361717.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04020936.hg.1 |
ENST00000438746.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04020936.hg.1 |
NM_052864.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04020945.hg.1 |
TC04001476.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04020945.hg.1 |
ENST00000361717.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04020945.hg.1 |
ENST00000438746.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04020945.hg.1 |
NM_052864.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011616.hg.1 |
ENST00000361717.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011616.hg.1 |
ENST00000361717.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011616.hg.1 |
ENST00000361717.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011616.hg.1 |
ENST00000361717.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011616.hg.1 |
ENST00000361717.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011616.hg.1 |
ENST00000361717.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011616.hg.1 |
ENST00000500655.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011616.hg.1 |
ENST00000500655.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011616.hg.1 |
ENST00000500655.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011616.hg.1 |
NM_052864.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011616.hg.1 |
NM_052864.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011616.hg.1 |
NM_052864.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011616.hg.1 |
NM_052864.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011616.hg.1 |
NM_052864.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04020939.hg.1 |
TC04001476.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04020939.hg.1 |
ENST00000361717.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04020939.hg.1 |
ENST00000438746.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04020939.hg.1 |
ENST00000500655.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04020939.hg.1 |
NM_052864.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04020942.hg.1 |
TC04001476.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04020942.hg.1 |
ENST00000361717.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04020942.hg.1 |
ENST00000438746.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04020942.hg.1 |
ENST00000500655.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04020942.hg.1 |
NM_052864.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04020940.hg.1 |
TC04001476.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04020940.hg.1 |
ENST00000361717.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04020940.hg.1 |
ENST00000438746.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04020940.hg.1 |
ENST00000500655.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04020940.hg.1 |
NM_052864.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400179765.hg.1 |
ENST00000361717.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179765.hg.1 |
NM_052864.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400179765.hg.1 |
ENST00000361717.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179765.hg.1 |
NM_052864.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400179765.hg.1 |
TC0400011616.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400179766.hg.1 |
ENST00000361717.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179766.hg.1 |
ENST00000500655.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179766.hg.1 |
NM_052864.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400179766.hg.1 |
ENST00000361717.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179766.hg.1 |
ENST00000500655.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179766.hg.1 |
NM_052864.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400179766.hg.1 |
TC0400011616.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400179767.hg.1 |
ENST00000361717.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179767.hg.1 |
ENST00000500655.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179767.hg.1 |
NM_052864.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400179767.hg.1 |
ENST00000361717.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179767.hg.1 |
ENST00000500655.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179767.hg.1 |
NM_052864.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400179767.hg.1 |
TC0400011616.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400011616.hg.1 |
ENST00000361717.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011616.hg.1 |
ENST00000361717.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011616.hg.1 |
ENST00000361717.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011616.hg.1 |
ENST00000361717.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011616.hg.1 |
ENST00000361717.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011616.hg.1 |
ENST00000361717.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011616.hg.1 |
ENST00000500655.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011616.hg.1 |
ENST00000500655.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011616.hg.1 |
ENST00000500655.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011616.hg.1 |
NM_052864.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011616.hg.1 |
NM_052864.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011616.hg.1 |
NM_052864.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011616.hg.1 |
NM_052864.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011616.hg.1 |
NM_052864.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400179770.hg.1 |
ENST00000361717.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179770.hg.1 |
NM_052864.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400179770.hg.1 |
ENST00000361717.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179770.hg.1 |
NM_052864.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400179770.hg.1 |
TC0400011616.hg.1 |
Clariom D TC |