Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008113.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008113.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008113.hg.1 |
TC16000561.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000561566.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000561566.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000561566.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000561856.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000561856.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000561856.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000561931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000561931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000561931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000561931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000561931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000561931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000562000.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000562000.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000562000.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000562000.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000562000.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000562491.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000562491.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000562491.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000562491.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000562491.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000564180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000564180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000564180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000564180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000564180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000564180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000565037.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000565037.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000565852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000565852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000565852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000565852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000565852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000568361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000568361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000568361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000568361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000568361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
ENST00000568361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161640.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161640.hg.1 |
ENST00000561856.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161640.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161640.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161640.hg.1 |
ENST00000561856.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161640.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161640.hg.1 |
TC1600008214.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000561566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000561566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000561566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000561856.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000561856.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000561856.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000561931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000561931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000561931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000561931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000561931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000561931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000562000.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000562000.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000562000.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000562000.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000562000.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000562491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000562491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000562491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000562491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000562491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000564180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000564180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000564180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000564180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000564180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000564180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000565037.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000565037.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000565852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000565852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000565852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000565852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000565852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000568361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000568361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000568361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000568361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000568361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
ENST00000568361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008214.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161641.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161641.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161641.hg.1 |
ENST00000261778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600161641.hg.1 |
NM_024562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600161641.hg.1 |
TC1600008214.hg.1 |
Clariom D TC |