Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000376840.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000376840.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000376840.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000376840.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000376840.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000376840.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000376840.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000376840.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000376840.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000376840.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000376840.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000376840.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000376840.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000376840.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000376840.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000376840.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000376840.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000376840.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000376840.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000376840.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000376840.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000376840.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000376840.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000376840.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000409318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000409318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000409318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000409318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000409318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000409318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000409318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000409318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000409318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000409318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000409318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000409318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000409318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000409318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000409318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000409318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000409318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000409318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000409318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000409318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000409318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000409318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000409318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000409318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000409318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000409318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000473937.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000473937.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000473937.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000473937.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000473937.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000473937.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000481317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000481317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000481317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000481317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000485875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000485875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000485875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000487392.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000487392.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000487392.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000487392.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000494011.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000494011.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000494011.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
ENST00000494011.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
NM_001102426.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
NM_001102426.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
NM_001102426.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
NM_001102426.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
NM_001102426.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
NM_001102426.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
NM_001102426.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
NM_001102426.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
NM_001102426.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
NM_001102426.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
NM_001102426.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
NM_001102426.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
NM_001102426.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
NM_001102426.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
NM_001102426.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
NM_001102426.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
NM_001102426.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
NM_001102426.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
NM_001102426.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
NM_001102426.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
NM_001102426.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
NM_001102426.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
NM_001102426.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
NM_001102426.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
NM_001102426.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016717.hg.1 |
NM_001102426.2 |
Refseq |