Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFYVE26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015346.3 | NP_056161.2 | 27 - 28 | 5456 | 96 | |
XM_017021124.1 | XP_016876613.1 | 27 - 28 | 5475 | 96 | ||
XM_017021125.1 | XP_016876614.1 | 27 - 28 | 5475 | 96 | ||
XM_017021126.1 | XP_016876615.1 | 26 - 27 | 6089 | 96 | ||
XM_017021127.1 | XP_016876616.1 | 16 - 17 | 3171 | 96 | ||
XM_017021128.1 | XP_016876617.1 | 16 - 17 | 3024 | 96 | ||
GenBank mRNA | AB002319.1 | - | 12 - 13 | 2326 | 96 | |
AB425197.1 | - | 27 - 28 | 5444 | 96 | ||
BC144594.1 | - | 25 - 26 | 4760 | 96 | ||
BX538025.1 | - | 23 - 24 | 4949 | 96 | ||
BX648683.1 | - | 27 - 28 | 5454 | 96 | ||
CR749276.1 | - | 28 - 29 | 5391 | 96 | ||
CR749365.1 | - | 27 - 28 | 5455 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons |