Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDM6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080424.1 | NP_001073893.1 | 13 - 14 | 4051 | 82 | |
XM_005256549.3 | XP_005256606.1 | 14 - 15 | 4203 | 82 | ||
XM_005256550.4 | XP_005256607.1 | 13 - 14 | 4045 | 82 | ||
XM_005256551.3 | XP_005256608.1 | 13 - 14 | 4084 | 82 | ||
XM_005256552.4 | XP_005256609.1 | 13 - 14 | 5057 | 82 | ||
XM_006721483.3 | XP_006721546.1 | 14 - 15 | 4148 | 82 | ||
XM_011523750.2 | XP_011522052.1 | 14 - 15 | 5408 | 82 | ||
XM_011523752.2 | XP_011522054.1 | 13 - 14 | 4048 | 82 | ||
XM_017024381.1 | XP_016879870.1 | 13 - 14 | 5494 | 82 | ||
GenBank mRNA | AB002344.2 | - | 13 - 14 | 4056 | 82 | |
BC009994.1 | - | 3 - 4 | 680 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons |