Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUSC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014806.3 | NP_055621.1 | 1 - 2 | 218 | 108 | |
NR_052015.2 | 1 - 2 | 218 | 108 | |||
GenBank mRNA | AB002373.2 | - | 1 - 2 | 60 | 108 | |
BC150211.1 | - | 1 - 2 | 130 | 108 | ||
BC150262.1 | - | 1 - 2 | 71 | 9007205 | 108 | |
DB458531.2 | - | 1 - 2 | 162 | 108 | ||
DB460837.2 | - | 1 - 2 | 219 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons |