Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07000173.hg.1 |
TC07003315.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07000173.hg.1 |
ENST00000297445.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07000173.hg.1 |
ENST00000389574.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07000173.hg.1 |
ENST00000401592.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07000173.hg.1 |
ENST00000405266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07000173.hg.1 |
ENST00000413514.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07000173.hg.1 |
ENST00000425407.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07000173.hg.1 |
ENST00000452783.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07000173.hg.1 |
ENST00000456758.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07000173.hg.1 |
NM_001130965.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07000173.hg.1 |
NM_001171944.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07000173.hg.1 |
NM_025154.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700196107.hg.1 |
ENST00000389574.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196107.hg.1 |
ENST00000401592.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196107.hg.1 |
ENST00000405266.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196107.hg.1 |
ENST00000413171.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196107.hg.1 |
ENST00000425407.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196107.hg.1 |
ENST00000429178.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196107.hg.1 |
ENST00000433212.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196107.hg.1 |
ENST00000452783.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196107.hg.1 |
ENST00000475971.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196107.hg.1 |
NM_001130965.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700196107.hg.1 |
NM_001171944.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700196107.hg.1 |
NM_025154.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700196107.hg.1 |
ENST00000389574.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196107.hg.1 |
ENST00000401592.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196107.hg.1 |
ENST00000405266.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196107.hg.1 |
ENST00000413171.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196107.hg.1 |
ENST00000425407.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196107.hg.1 |
ENST00000429178.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196107.hg.1 |
ENST00000433212.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196107.hg.1 |
ENST00000452783.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196107.hg.1 |
ENST00000475971.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196107.hg.1 |
NM_001130965.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700196107.hg.1 |
NM_001171944.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700196107.hg.1 |
NM_025154.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700196107.hg.1 |
TC0700013323.hg.1 |
Clariom D TC |