Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130960.1 | NP_001124432.1 | 21 - 22 | 2926 | 122 | |
NM_001130961.1 | NP_001124433.1 | 21 - 22 | 2926 | 122 | ||
XM_005247238.1 | XP_005247295.1 | 22 - 23 | 2975 | 122 | ||
XM_011512560.2 | XP_011510862.1 | 22 - 23 | 2984 | 122 | ||
XM_011512561.2 | XP_011510863.1 | 22 - 23 | 3116 | 122 | ||
XM_011512562.2 | XP_011510864.1 | 23 - 24 | 3733 | 122 | ||
XM_011512565.2 | XP_011510867.1 | 21 - 22 | 3447 | 122 | ||
XM_011512566.2 | XP_011510868.1 | 21 - 22 | 3088 | 122 | ||
XM_011512567.1 | XP_011510869.1 | 22 - 23 | 2975 | 122 | ||
XM_017005923.1 | XP_016861412.1 | 22 - 23 | 2975 | 122 | ||
XM_017005925.1 | XP_016861414.1 | 21 - 22 | 2903 | 122 | ||
XM_017005926.1 | XP_016861415.1 | 21 - 22 | 2874 | 122 | ||
XM_017005927.1 | XP_016861416.1 | 22 - 23 | 2975 | 122 | ||
XM_017005928.1 | XP_016861417.1 | 22 - 23 | 2975 | 122 | ||
XM_017005929.1 | XP_016861418.1 | 21 - 22 | 3026 | 122 | ||
GenBank mRNA | AB028992.1 | - | 17 - 18 | 2004 | 122 | |
AY691170.1 | - | 21 - 22 | 2649 | 122 | ||
AY691171.1 | - | 21 - 22 | 2649 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Probe spans exons |