Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPIRE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128626.1 | NP_001122098.1 | 12 - 13 | 1690 | 77 | |
NM_001128627.1 | NP_001122099.1 | 11 - 12 | 1346 | 77 | ||
NM_020148.2 | NP_064533.3 | 11 - 12 | 1648 | 77 | ||
GenBank mRNA | AB032961.2 | - | 12 - 13 | 1742 | 77 | |
AJ277587.1 | - | 9 - 10 | 1235 | 77 | ||
AK290180.1 | - | 9 - 10 | 1206 | 77 | ||
AK301711.1 | - | 10 - 11 | 1115 | 77 | ||
AL833817.1 | - | 12 - 13 | 1326 | 77 | ||
BC115005.1 | - | 11 - 12 | 1473 | 7782242 | 77 | |
BC125206.1 | - | 10 - 11 | 1204 | 40123468 | 77 | |
BC125207.1 | - | 9 - 10 | 1162 | 40123469 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |