Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029884.1 | NP_001025055.1 | 12 - 13 | 1630 | 71 | |
XM_005267064.3 | XP_005267121.1 | 11 - 12 | 3709 | 71 | ||
XM_005267065.3 | XP_005267122.1 | 13 - 14 | 1656 | 71 | ||
XM_005267066.2 | XP_005267123.1 | 11 - 12 | 2513 | 71 | ||
XM_006715521.1 | XP_006715584.1 | 13 - 14 | 1883 | 71 | ||
XM_006715522.3 | XP_006715585.1 | 11 - 12 | 3750 | 71 | ||
XM_011535981.2 | XP_011534283.1 | 11 - 12 | 4506 | 71 | ||
XM_011535982.2 | XP_011534284.1 | 11 - 12 | 4506 | 71 | ||
XM_011535983.2 | XP_011534285.1 | 12 - 13 | 2931 | 71 | ||
XM_017011102.1 | XP_016866591.1 | 11 - 12 | 2515 | 71 | ||
GenBank mRNA | AB033035.1 | - | 5 - 6 | 503 | 71 | |
AJ420468.1 | - | 9 - 10 | 868 | 71 | ||
AK056300.1 | - | 11 - 12 | 1529 | 71 | ||
AK293288.1 | - | 11 - 12 | 1561 | 71 | ||
AK294798.1 | - | 11 - 12 | 1684 | 71 | ||
AK307631.1 | - | 11 - 12 | 1583 | 71 | ||
BC089428.1 | - | 7 - 8 | 741 | 71 | ||
BC140864.1 | - | 10 - 11 | 1367 | 9021732 | 71 | |
BC151134.1 | - | 10 - 11 | 1367 | 9057034 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |