Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTSL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301110.1 | NP_001288039.1 | 17 - 18 | 2343 | 93 | |
NM_207517.2 | NP_997400.2 | 17 - 18 | 2343 | 93 | ||
XM_011521822.2 | XP_011520124.1 | 18 - 19 | 2415 | 93 | ||
XM_011521823.2 | XP_011520125.1 | 18 - 19 | 2415 | 93 | ||
XM_011521824.2 | XP_011520126.1 | 18 - 19 | 2415 | 93 | ||
XM_011521825.2 | XP_011520127.1 | 18 - 19 | 2415 | 93 | ||
XM_017022434.1 | XP_016877923.1 | 18 - 19 | 2415 | 93 | ||
XM_017022435.1 | XP_016877924.1 | 17 - 18 | 2211 | 93 | ||
GenBank mRNA | AB033059.1 | - | 1 - 2 | 117 | 93 | |
AF237652.1 | - | 17 - 18 | 2343 | 93 | ||
AK308845.1 | - | 17 - 18 | 2334 | 93 | ||
BC128389.1 | - | 17 - 18 | 2341 | 40125494 | 93 | |
BC128390.1 | - | 17 - 18 | 2341 | 40125501 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons |