Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
ENST00000289877.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
ENST00000289877.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
ENST00000289877.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
ENST00000289877.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
ENST00000289877.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
ENST00000289877.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
ENST00000289877.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
ENST00000289877.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
ENST00000289877.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
ENST00000289877.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
ENST00000471889.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
ENST00000471889.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
ENST00000471889.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
ENST00000471889.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
ENST00000471889.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
ENST00000471889.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
ENST00000471889.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
ENST00000471889.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
ENST00000471889.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
ENST00000471889.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
ENST00000471889.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
ENST00000471889.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
ENST00000481265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
ENST00000481265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
ENST00000481265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
ENST00000497660.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
ENST00000497660.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
ENST00000497660.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
ENST00000497660.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
ENST00000497660.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
ENST00000497660.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
ENST00000497660.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
ENST00000497660.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
NM_001080397.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
NM_001080397.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
NM_001080397.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
NM_001080397.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
NM_001080397.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
NM_001080397.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
NM_001080397.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
NM_001080397.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
NM_001080397.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
NM_001080397.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006739.hg.1 |
NM_001080397.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001689.hg.1 |
TC01000104.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001689.hg.1 |
ENST00000289877.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001689.hg.1 |
ENST00000377479.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001689.hg.1 |
ENST00000471889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001689.hg.1 |
NM_001080397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100146695.hg.1 |
ENST00000289877.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100146695.hg.1 |
ENST00000471889.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100146695.hg.1 |
ENST00000497660.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100146695.hg.1 |
NM_001080397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100146695.hg.1 |
ENST00000289877.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100146695.hg.1 |
ENST00000471889.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100146695.hg.1 |
ENST00000497660.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100146695.hg.1 |
NM_001080397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100146695.hg.1 |
TC0100006739.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
ENST00000289877.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
ENST00000289877.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
ENST00000289877.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
ENST00000289877.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
ENST00000289877.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
ENST00000289877.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
ENST00000289877.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
ENST00000289877.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
ENST00000289877.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
ENST00000289877.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
ENST00000471889.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
ENST00000471889.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
ENST00000471889.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
ENST00000471889.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
ENST00000471889.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
ENST00000471889.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
ENST00000471889.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
ENST00000471889.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
ENST00000471889.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
ENST00000471889.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
ENST00000471889.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
ENST00000471889.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
ENST00000481265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
ENST00000481265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
ENST00000481265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
ENST00000497660.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
ENST00000497660.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
ENST00000497660.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
ENST00000497660.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
ENST00000497660.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
ENST00000497660.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
ENST00000497660.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
ENST00000497660.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
NM_001080397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
NM_001080397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
NM_001080397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
NM_001080397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
NM_001080397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
NM_001080397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
NM_001080397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
NM_001080397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
NM_001080397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
NM_001080397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006739.hg.1 |
NM_001080397.2 |
Refseq |