Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
ENST00000360565.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
ENST00000360565.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
ENST00000360565.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
ENST00000360565.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
ENST00000360565.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
ENST00000360565.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
ENST00000360565.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
ENST00000360565.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
ENST00000360565.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
ENST00000360565.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
ENST00000360565.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
ENST00000360565.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
ENST00000360565.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
ENST00000360565.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
ENST00000595242.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
ENST00000595242.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
ENST00000595242.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
ENST00000595242.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
ENST00000595242.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
ENST00000595242.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
ENST00000598359.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
ENST00000598359.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
ENST00000598359.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
ENST00000598359.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
ENST00000598359.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
ENST00000598359.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
ENST00000598359.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
ENST00000598359.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
ENST00000601038.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
ENST00000601038.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
ENST00000601038.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
ENST00000601038.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
ENST00000601038.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
ENST00000601038.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
NM_021228.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
NM_021228.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
NM_021228.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
NM_021228.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
NM_021228.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
NM_021228.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
NM_021228.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
NM_021228.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
NM_021228.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
NM_021228.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
NM_021228.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
NM_021228.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
NM_021228.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008549.hg.1 |
NM_021228.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010385.hg.1 |
ENST00000360565.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010385.hg.1 |
NM_021228.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010385.hg.1 |
TC19000742.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900166268.hg.1 |
ENST00000360565.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900166268.hg.1 |
NM_021228.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900166268.hg.1 |
TC1900008549.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900166268.hg.1 |
ENST00000360565.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900166268.hg.1 |
NM_021228.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
ENST00000360565.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
ENST00000360565.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
ENST00000360565.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
ENST00000360565.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
ENST00000360565.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
ENST00000360565.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
ENST00000360565.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
ENST00000360565.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
ENST00000360565.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
ENST00000360565.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
ENST00000360565.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
ENST00000360565.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
ENST00000360565.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
ENST00000360565.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
ENST00000595242.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
ENST00000595242.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
ENST00000595242.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
ENST00000595242.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
ENST00000595242.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
ENST00000595242.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
ENST00000598359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
ENST00000598359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
ENST00000598359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
ENST00000598359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
ENST00000598359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
ENST00000598359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
ENST00000598359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
ENST00000598359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
ENST00000601038.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
ENST00000601038.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
ENST00000601038.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
ENST00000601038.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
ENST00000601038.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
ENST00000601038.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
NM_021228.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
NM_021228.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
NM_021228.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
NM_021228.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
NM_021228.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
NM_021228.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
NM_021228.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
NM_021228.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
NM_021228.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
NM_021228.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
NM_021228.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
NM_021228.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
NM_021228.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008549.hg.1 |
NM_021228.2 |
Refseq |