Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009163.hg.1 |
TC04000632.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009163.hg.1 |
ENST00000394339.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009163.hg.1 |
NM_005841.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009165.hg.1 |
TC04000632.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009165.hg.1 |
ENST00000339241.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009165.hg.1 |
ENST00000394339.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009165.hg.1 |
NM_005841.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009165.hg.1 |
NM_199327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000339241.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000339241.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000339241.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000339241.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000339241.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000339241.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000339241.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000394339.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000394339.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000394339.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000394339.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000394339.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000394339.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000394339.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000505319.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000505319.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000505319.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000505319.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000505319.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000507703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000507703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000507703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000507703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000507703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000507703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000507703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000508849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000508849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000508849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000515726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000515726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000515726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000515726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000610581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000610581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000610581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000610581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000610581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000610581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000610581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000610581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000622283.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000622283.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000622283.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000622283.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000622283.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000622283.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
ENST00000622283.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
NM_001258038.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
NM_001258038.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
NM_001258038.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
NM_001258038.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
NM_001258038.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
NM_001258038.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
NM_001258038.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
NM_001258038.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
NM_001258039.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
NM_001258039.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
NM_001258039.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
NM_001258039.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
NM_001258039.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
NM_001258039.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
NM_001258039.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
NM_005841.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
NM_005841.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
NM_005841.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
NM_005841.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
NM_005841.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
NM_005841.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
NM_005841.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
NM_199327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
NM_199327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
NM_199327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
NM_199327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
NM_199327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
NM_199327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008628.hg.1 |
NM_199327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009168.hg.1 |
TC04000632.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009168.hg.1 |
ENST00000339241.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009168.hg.1 |
ENST00000394339.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009168.hg.1 |
NM_005841.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009168.hg.1 |
NM_199327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009166.hg.1 |
TC04000632.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009166.hg.1 |
ENST00000339241.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009166.hg.1 |
ENST00000394339.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009166.hg.1 |
NM_005841.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009166.hg.1 |
NM_199327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009167.hg.1 |
TC04000632.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009167.hg.1 |
ENST00000339241.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009167.hg.1 |
ENST00000394339.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009167.hg.1 |
NM_005841.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009167.hg.1 |
NM_199327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009169.hg.1 |
TC04000632.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009169.hg.1 |
ENST00000339241.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009169.hg.1 |
ENST00000394339.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009169.hg.1 |
NM_005841.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009169.hg.1 |
NM_199327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000339241.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000339241.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000339241.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000339241.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000339241.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000339241.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000339241.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000394339.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000394339.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000394339.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000394339.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000394339.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000394339.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000394339.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000505319.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000505319.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000505319.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000505319.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000505319.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000507703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000507703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000507703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000507703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000507703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000507703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000507703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000508849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000508849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000508849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000515726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000515726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000515726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000515726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000610581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000610581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000610581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000610581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000610581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000610581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000610581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000610581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000622283.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000622283.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000622283.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000622283.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000622283.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000622283.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
ENST00000622283.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
NM_001258038.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
NM_001258038.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
NM_001258038.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
NM_001258038.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
NM_001258038.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
NM_001258038.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
NM_001258038.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
NM_001258038.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
NM_001258039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
NM_001258039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
NM_001258039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
NM_001258039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
NM_001258039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
NM_001258039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
NM_001258039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
NM_005841.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
NM_005841.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
NM_005841.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
NM_005841.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
NM_005841.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
NM_005841.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
NM_005841.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
NM_199327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
NM_199327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
NM_199327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
NM_199327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
NM_199327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
NM_199327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008628.hg.1 |
NM_199327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400159305.hg.1 |
ENST00000394339.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400159305.hg.1 |
NM_005841.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400159305.hg.1 |
ENST00000394339.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400159305.hg.1 |
NM_005841.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400159305.hg.1 |
TC0400008628.hg.1 |
Clariom D TC |