Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000471328.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000471328.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000477004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000477004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000477004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000477004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000512382.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000512382.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000512382.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000512382.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000515795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000515795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000515795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000515795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000515795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000515795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000515795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000515795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000515795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07000029.hg.1 |
TC07000004.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07000029.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07000029.hg.1 |
NM_020223.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000471328.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000471328.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000477004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000477004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000477004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000477004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000512382.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000512382.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000512382.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000512382.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000515795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000515795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000515795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000515795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000515795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000515795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000515795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000515795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000515795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700144126.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700144126.hg.1 |
ENST00000515795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700144126.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700144126.hg.1 |
ENST00000515795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700144126.hg.1 |
TC0700006445.hg.1 |
Clariom D TC |