Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FANCI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113378.1 | NP_001106849.1 | 30 - 31 | 3343 | 97 | |
NM_018193.2 | NP_060663.2 | 29 - 30 | 3163 | 97 | ||
XM_011521756.2 | XP_011520058.1 | 30 - 31 | 3367 | 97 | ||
XM_011521757.2 | XP_011520059.1 | 30 - 31 | 3363 | 97 | ||
XM_011521764.2 | XP_011520066.1 | 29 - 30 | 3187 | 97 | ||
XM_011521767.2 | XP_011520069.1 | 30 - 31 | 3337 | 97 | ||
GenBank mRNA | AB058697.1 | - | 16 - 17 | 3267 | 97 | |
AK001581.1 | - | 23 - 24 | 2552 | 97 | ||
AK027564.1 | - | 18 - 19 | 2050 | 97 | ||
AK055176.1 | - | 29 - 30 | 3122 | 97 | ||
BC021859.1 | - | 7 - 8 | 685 | 97 | ||
BC140769.1 | - | 30 - 31 | 3300 | 9021637 | 97 | |
BC144483.1 | - | 30 - 31 | 3300 | 9053010 | 97 | |
EF469766.1 | - | 29 - 30 | 3253 | 97 | ||
EF567077.1 | - | 29 - 30 | 3253 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |