Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
ENST00000468890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
ENST00000468890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
ENST00000468890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
ENST00000468890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
NM_198514.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
NM_198514.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
NM_198514.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
NM_198514.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
NM_198514.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
NM_198514.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
NM_198514.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
NM_198514.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
NM_198514.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
NM_198514.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
NM_198514.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
NM_198514.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
NM_198514.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
NM_198514.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
NM_198514.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
NM_198514.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008961.hg.1 |
NM_198514.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009992.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009992.hg.1 |
NM_198514.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009992.hg.1 |
TC10000828.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000161310.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161310.hg.1 |
NM_198514.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161310.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161310.hg.1 |
NM_198514.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161310.hg.1 |
TC1000008961.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
ENST00000369301.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
ENST00000468890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
ENST00000468890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
ENST00000468890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
ENST00000468890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
NM_198514.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
NM_198514.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
NM_198514.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
NM_198514.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
NM_198514.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
NM_198514.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
NM_198514.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
NM_198514.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
NM_198514.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
NM_198514.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
NM_198514.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
NM_198514.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
NM_198514.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
NM_198514.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
NM_198514.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
NM_198514.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008961.hg.1 |
NM_198514.3 |
Refseq |