Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14001184.hg.1 |
ENST00000267396.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14001184.hg.1 |
ENST00000536884.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14001184.hg.1 |
NM_173527.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14001184.hg.1 |
TC14000136.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14001185.hg.1 |
ENST00000267396.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14001185.hg.1 |
ENST00000536884.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14001185.hg.1 |
NM_173527.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14001185.hg.1 |
TC14000136.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006664.hg.1 |
ENST00000267396.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006664.hg.1 |
ENST00000267396.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006664.hg.1 |
ENST00000267396.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006664.hg.1 |
ENST00000267396.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006664.hg.1 |
ENST00000267396.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006664.hg.1 |
ENST00000267396.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006664.hg.1 |
ENST00000267396.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006664.hg.1 |
ENST00000267396.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006664.hg.1 |
ENST00000267396.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006664.hg.1 |
ENST00000536884.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006664.hg.1 |
ENST00000536884.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006664.hg.1 |
ENST00000536884.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006664.hg.1 |
ENST00000536884.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006664.hg.1 |
ENST00000536884.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006664.hg.1 |
ENST00000536884.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006664.hg.1 |
NM_173527.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006664.hg.1 |
NM_173527.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006664.hg.1 |
NM_173527.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006664.hg.1 |
NM_173527.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006664.hg.1 |
NM_173527.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006664.hg.1 |
NM_173527.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006664.hg.1 |
NM_173527.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006664.hg.1 |
NM_173527.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400145574.hg.1 |
ENST00000267396.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400145574.hg.1 |
ENST00000536884.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400145574.hg.1 |
NM_173527.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400145574.hg.1 |
ENST00000267396.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400145574.hg.1 |
ENST00000536884.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400145574.hg.1 |
NM_173527.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400145574.hg.1 |
TC1400006664.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400145573.hg.1 |
ENST00000267396.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400145573.hg.1 |
ENST00000536884.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400145573.hg.1 |
NM_173527.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400145573.hg.1 |
ENST00000267396.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400145573.hg.1 |
ENST00000536884.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400145573.hg.1 |
NM_173527.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400145573.hg.1 |
TC1400006664.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400006664.hg.1 |
ENST00000267396.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006664.hg.1 |
ENST00000267396.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006664.hg.1 |
ENST00000267396.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006664.hg.1 |
ENST00000267396.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006664.hg.1 |
ENST00000267396.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006664.hg.1 |
ENST00000267396.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006664.hg.1 |
ENST00000267396.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006664.hg.1 |
ENST00000267396.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006664.hg.1 |
ENST00000267396.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006664.hg.1 |
ENST00000536884.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006664.hg.1 |
ENST00000536884.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006664.hg.1 |
ENST00000536884.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006664.hg.1 |
ENST00000536884.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006664.hg.1 |
ENST00000536884.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006664.hg.1 |
ENST00000536884.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006664.hg.1 |
NM_173527.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006664.hg.1 |
NM_173527.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006664.hg.1 |
NM_173527.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006664.hg.1 |
NM_173527.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006664.hg.1 |
NM_173527.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006664.hg.1 |
NM_173527.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006664.hg.1 |
NM_173527.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006664.hg.1 |
NM_173527.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400145575.hg.1 |
ENST00000267396.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400145575.hg.1 |
ENST00000536884.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400145575.hg.1 |
NM_173527.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400145575.hg.1 |
ENST00000267396.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400145575.hg.1 |
ENST00000536884.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400145575.hg.1 |
NM_173527.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400145575.hg.1 |
TC1400006664.hg.1 |
Clariom D TC |