Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
ENST00000237172.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
ENST00000237172.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
ENST00000237172.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
ENST00000237172.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
ENST00000237172.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
ENST00000237172.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
ENST00000237172.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
ENST00000237172.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
ENST00000237172.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
ENST00000237172.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
ENST00000370020.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
ENST00000370020.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
ENST00000370020.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
ENST00000370020.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
ENST00000370020.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
ENST00000370020.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
ENST00000370020.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
ENST00000370020.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
ENST00000498523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
ENST00000498523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
ENST00000498523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
ENST00000498523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
ENST00000498523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
ENST00000498523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
NM_001289987.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
NM_001289987.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
NM_001289987.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
NM_001289987.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
NM_001289987.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
NM_001289987.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
NM_001289987.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
NM_001289987.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
NM_001289987.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
NM_001289987.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
NM_001289987.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
NM_001289987.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
NM_001300866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
NM_001300866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
NM_001300866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
NM_001300866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
NM_001300866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
NM_001300866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
NM_001300866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
NM_001300866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
NM_001300866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
NM_001300866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
NM_015687.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
NM_015687.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
NM_015687.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
NM_015687.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
NM_015687.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
NM_015687.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
NM_015687.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
NM_015687.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
NM_015687.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
NM_015687.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
NM_015687.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
NR_110608.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
NR_110608.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
NR_110608.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014316.hg.1 |
NR_110608.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022949.hg.1 |
TC06001882.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022949.hg.1 |
ENST00000237172.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022949.hg.1 |
ENST00000370020.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022949.hg.1 |
ENST00000393004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022949.hg.1 |
ENST00000498523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022949.hg.1 |
NM_015687.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202186.hg.1 |
ENST00000237172.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202186.hg.1 |
ENST00000370020.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202186.hg.1 |
ENST00000498523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202186.hg.1 |
NM_001289987.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202186.hg.1 |
NM_001300866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202186.hg.1 |
NM_015687.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202186.hg.1 |
ENST00000237172.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202186.hg.1 |
ENST00000370020.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202186.hg.1 |
ENST00000498523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202186.hg.1 |
NM_001289987.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202186.hg.1 |
NM_001300866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202186.hg.1 |
NM_015687.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202186.hg.1 |
TC0600014316.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
ENST00000237172.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
ENST00000237172.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
ENST00000237172.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
ENST00000237172.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
ENST00000237172.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
ENST00000237172.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
ENST00000237172.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
ENST00000237172.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
ENST00000237172.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
ENST00000237172.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
ENST00000370020.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
ENST00000370020.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
ENST00000370020.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
ENST00000370020.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
ENST00000370020.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
ENST00000370020.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
ENST00000370020.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
ENST00000370020.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
ENST00000498523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
ENST00000498523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
ENST00000498523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
ENST00000498523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
ENST00000498523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
ENST00000498523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
NM_001289987.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
NM_001289987.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
NM_001289987.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
NM_001289987.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
NM_001289987.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
NM_001289987.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
NM_001289987.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
NM_001289987.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
NM_001289987.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
NM_001289987.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
NM_001289987.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
NM_001289987.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
NM_001300866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
NM_001300866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
NM_001300866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
NM_001300866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
NM_001300866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
NM_001300866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
NM_001300866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
NM_001300866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
NM_001300866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
NM_001300866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
NM_015687.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
NM_015687.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
NM_015687.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
NM_015687.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
NM_015687.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
NM_015687.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
NM_015687.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
NM_015687.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
NM_015687.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
NM_015687.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
NM_015687.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
NR_110608.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
NR_110608.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
NR_110608.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014316.hg.1 |
NR_110608.2 |
Refseq |