Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05006473.hg.1 |
TC05000458.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05006473.hg.1 |
ENST00000380009.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05006473.hg.1 |
ENST00000537857.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05006473.hg.1 |
NM_198150.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05006475.hg.1 |
TC05000458.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05006475.hg.1 |
ENST00000380009.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05006475.hg.1 |
ENST00000537857.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05006475.hg.1 |
NM_198150.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000380009.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000380009.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000380009.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000380009.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000380009.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000380009.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000380009.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000380009.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000380009.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000380009.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000380009.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000380009.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000380009.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000380009.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000380009.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000504763.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000504763.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000504763.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000504763.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000504873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000504873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000504873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000504873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000504873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000504873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000504873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000504873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000504873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000504873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000504873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000504873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000504873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000512106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000512106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000512106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000512106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000513814.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000513814.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000513814.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000513814.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000513814.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000513814.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000513814.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000513814.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000513814.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000513814.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000513814.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000513814.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000513814.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
ENST00000513814.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
NM_198150.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
NM_198150.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
NM_198150.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
NM_198150.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
NM_198150.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
NM_198150.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
NM_198150.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
NM_198150.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
NM_198150.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
NM_198150.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
NM_198150.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
NM_198150.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
NM_198150.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
NM_198150.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008121.hg.1 |
NM_198150.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500155559.hg.1 |
ENST00000380009.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155559.hg.1 |
ENST00000504873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155559.hg.1 |
ENST00000512106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155559.hg.1 |
ENST00000513814.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155559.hg.1 |
NM_198150.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500155559.hg.1 |
ENST00000380009.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155559.hg.1 |
ENST00000504873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155559.hg.1 |
ENST00000512106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155559.hg.1 |
ENST00000513814.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500155559.hg.1 |
NM_198150.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500155559.hg.1 |
TC0500008121.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000380009.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000380009.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000380009.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000380009.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000380009.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000380009.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000380009.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000380009.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000380009.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000380009.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000380009.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000380009.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000380009.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000380009.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000380009.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000504763.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000504763.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000504763.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000504763.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000504873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000504873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000504873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000504873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000504873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000504873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000504873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000504873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000504873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000504873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000504873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000504873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000504873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000512106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000512106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000512106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000512106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000513814.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000513814.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000513814.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000513814.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000513814.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000513814.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000513814.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000513814.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000513814.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000513814.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000513814.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000513814.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000513814.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
ENST00000513814.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
NM_198150.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
NM_198150.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
NM_198150.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
NM_198150.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
NM_198150.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
NM_198150.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
NM_198150.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
NM_198150.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
NM_198150.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
NM_198150.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
NM_198150.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
NM_198150.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
NM_198150.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
NM_198150.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008121.hg.1 |
NM_198150.2 |
Refseq |