Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008273.hg.1 |
ENST00000326624.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008273.hg.1 |
ENST00000326624.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008273.hg.1 |
ENST00000326624.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008273.hg.1 |
ENST00000557385.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008273.hg.1 |
ENST00000557385.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008273.hg.1 |
ENST00000557385.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008273.hg.1 |
NM_001184742.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008273.hg.1 |
NM_001184742.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008273.hg.1 |
NM_001184742.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008273.hg.1 |
NM_001184742.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008273.hg.1 |
NM_006777.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008273.hg.1 |
NM_006777.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008273.hg.1 |
NM_006777.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007801.hg.1 |
TC0X000593.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007801.hg.1 |
ENST00000326624.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007801.hg.1 |
ENST00000540105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007801.hg.1 |
ENST00000557385.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007801.hg.1 |
NM_001184742.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007801.hg.1 |
NM_006777.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007804.hg.1 |
TC0X000593.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007804.hg.1 |
ENST00000326624.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007804.hg.1 |
ENST00000540105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007804.hg.1 |
ENST00000557385.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007804.hg.1 |
NM_001184742.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007804.hg.1 |
NM_006777.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007806.hg.1 |
TC0X000593.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007806.hg.1 |
ENST00000326624.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007806.hg.1 |
ENST00000540105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007806.hg.1 |
ENST00000557385.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007806.hg.1 |
NM_001184742.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007806.hg.1 |
NM_006777.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008273.hg.1 |
ENST00000326624.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008273.hg.1 |
ENST00000326624.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008273.hg.1 |
ENST00000326624.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008273.hg.1 |
ENST00000557385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008273.hg.1 |
ENST00000557385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008273.hg.1 |
ENST00000557385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008273.hg.1 |
NM_001184742.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008273.hg.1 |
NM_001184742.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008273.hg.1 |
NM_001184742.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008273.hg.1 |
NM_001184742.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008273.hg.1 |
NM_006777.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008273.hg.1 |
NM_006777.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008273.hg.1 |
NM_006777.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00156522.hg.1 |
ENST00000326624.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156522.hg.1 |
ENST00000557385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156522.hg.1 |
NM_001184742.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00156522.hg.1 |
NM_006777.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00156522.hg.1 |
ENST00000326624.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156522.hg.1 |
ENST00000557385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156522.hg.1 |
NM_001184742.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00156522.hg.1 |
NM_006777.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00156522.hg.1 |
TC0X00008273.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00156525.hg.1 |
ENST00000326624.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156525.hg.1 |
ENST00000557385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156525.hg.1 |
NM_001184742.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00156525.hg.1 |
NM_006777.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00156525.hg.1 |
ENST00000326624.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156525.hg.1 |
ENST00000557385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156525.hg.1 |
NM_001184742.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00156525.hg.1 |
NM_006777.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00156525.hg.1 |
TC0X00008273.hg.1 |
Clariom D TC |