Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023774.hg.1 |
TC06001940.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023774.hg.1 |
ENST00000435041.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023774.hg.1 |
ENST00000536477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023774.hg.1 |
NM_016021.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186425.hg.1 |
ENST00000435041.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186425.hg.1 |
NM_016021.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186425.hg.1 |
ENST00000435041.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186425.hg.1 |
NM_016021.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186425.hg.1 |
TC0600012542.hg.1 |
Clariom D TC |