Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STRIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270768.1 | NP_001257697.1 | 18 - 19 | 1845 | 78 | |
NM_033088.3 | NP_149079.2 | 18 - 19 | 1998 | 78 | ||
NR_073071.1 | 19 - 20 | 2112 | 78 | |||
XM_006710995.2 | XP_006711058.1 | 11 - 12 | 1101 | 78 | ||
XM_017002629.1 | XP_016858118.1 | 16 - 17 | 1769 | 78 | ||
GenBank mRNA | AB051548.2 | - | 18 - 19 | 1963 | 78 | |
AK027649.1 | - | 18 - 19 | 1845 | 78 | ||
AK098023.1 | - | 1 - 2 | 734 | 78 | ||
AK125054.1 | - | 9 - 10 | 2771 | 78 | ||
AK291059.1 | - | 18 - 19 | 1972 | 78 | ||
AL834196.1 | - | 19 - 20 | 2081 | 78 | ||
BC019919.1 | - | 9 - 10 | 791 | 78 | ||
BC032644.1 | - | 3 - 4 | 321 | 78 | ||
BC094786.1 | - | 18 - 19 | 1979 | 30342169 | 78 | |
BC119814.1 | - | 18 - 19 | 1980 | 40108348 | 78 | |
BC121793.1 | - | 18 - 19 | 1980 | 40108345 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |