Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012624.hg.1 |
ENST00000390649.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012624.hg.1 |
NM_153447.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012624.hg.1 |
TC19000915.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012625.hg.1 |
ENST00000390649.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012625.hg.1 |
NM_153447.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012625.hg.1 |
TC19000915.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
ENST00000390649.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
ENST00000390649.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
ENST00000390649.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
ENST00000390649.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
ENST00000390649.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
ENST00000390649.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
ENST00000390649.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
ENST00000390649.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
ENST00000390649.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
ENST00000390649.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
ENST00000390649.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
ENST00000390649.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
ENST00000390649.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
ENST00000390649.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
ENST00000390649.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
ENST00000390649.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
ENST00000597673.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
ENST00000597673.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
ENST00000597673.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
ENST00000597673.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
ENST00000597673.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
ENST00000621651.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
ENST00000621651.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
ENST00000621651.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
ENST00000621651.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
ENST00000621651.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
ENST00000621651.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
ENST00000621651.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
ENST00000621651.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
ENST00000621651.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
ENST00000621651.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
ENST00000621651.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
ENST00000621651.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
ENST00000621651.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
ENST00000621651.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
ENST00000621651.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
ENST00000621651.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
NM_153447.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
NM_153447.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
NM_153447.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
NM_153447.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
NM_153447.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
NM_153447.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
NM_153447.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
NM_153447.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
NM_153447.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
NM_153447.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
NM_153447.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
NM_153447.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
NM_153447.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
NM_153447.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
NM_153447.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008937.hg.1 |
NM_153447.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900170170.hg.1 |
ENST00000390649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170170.hg.1 |
ENST00000621651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170170.hg.1 |
NM_153447.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900170170.hg.1 |
TC1900008937.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900170170.hg.1 |
ENST00000390649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170170.hg.1 |
ENST00000621651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170170.hg.1 |
NM_153447.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
ENST00000390649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
ENST00000390649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
ENST00000390649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
ENST00000390649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
ENST00000390649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
ENST00000390649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
ENST00000390649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
ENST00000390649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
ENST00000390649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
ENST00000390649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
ENST00000390649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
ENST00000390649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
ENST00000390649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
ENST00000390649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
ENST00000390649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
ENST00000390649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
ENST00000597673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
ENST00000597673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
ENST00000597673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
ENST00000597673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
ENST00000597673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
ENST00000621651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
ENST00000621651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
ENST00000621651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
ENST00000621651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
ENST00000621651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
ENST00000621651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
ENST00000621651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
ENST00000621651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
ENST00000621651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
ENST00000621651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
ENST00000621651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
ENST00000621651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
ENST00000621651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
ENST00000621651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
ENST00000621651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
ENST00000621651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
NM_153447.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
NM_153447.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
NM_153447.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
NM_153447.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
NM_153447.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
NM_153447.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
NM_153447.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
NM_153447.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
NM_153447.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
NM_153447.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
NM_153447.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
NM_153447.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
NM_153447.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
NM_153447.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
NM_153447.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008937.hg.1 |
NM_153447.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900170169.hg.1 |
ENST00000390649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170169.hg.1 |
ENST00000621651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170169.hg.1 |
NM_153447.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900170169.hg.1 |
TC1900008937.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900170169.hg.1 |
ENST00000390649.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170169.hg.1 |
ENST00000621651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170169.hg.1 |
NM_153447.4 |
Refseq |