Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047138.hg.1 |
TC01003020.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047138.hg.1 |
ENST00000261448.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047138.hg.1 |
ENST00000446755.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047138.hg.1 |
NM_001232.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015378.hg.1 |
ENST00000261448.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015378.hg.1 |
ENST00000261448.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015378.hg.1 |
ENST00000261448.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015378.hg.1 |
ENST00000261448.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015378.hg.1 |
ENST00000261448.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015378.hg.1 |
ENST00000261448.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015378.hg.1 |
ENST00000261448.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015378.hg.1 |
ENST00000261448.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015378.hg.1 |
ENST00000261448.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015378.hg.1 |
ENST00000261448.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015378.hg.1 |
ENST00000261448.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015378.hg.1 |
ENST00000261448.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015378.hg.1 |
ENST00000261448.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015378.hg.1 |
ENST00000261448.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015378.hg.1 |
ENST00000261448.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015378.hg.1 |
ENST00000261448.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015378.hg.1 |
ENST00000488931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015378.hg.1 |
ENST00000488931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015378.hg.1 |
ENST00000488931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015378.hg.1 |
NM_001232.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015378.hg.1 |
NM_001232.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015378.hg.1 |
NM_001232.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015378.hg.1 |
NM_001232.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015378.hg.1 |
NM_001232.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015378.hg.1 |
NM_001232.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015378.hg.1 |
NM_001232.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015378.hg.1 |
NM_001232.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015378.hg.1 |
NM_001232.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015378.hg.1 |
NM_001232.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015378.hg.1 |
NM_001232.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015378.hg.1 |
NM_001232.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015378.hg.1 |
NM_001232.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015378.hg.1 |
NM_001232.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015378.hg.1 |
NM_001232.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015378.hg.1 |
NM_001232.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100214918.hg.1 |
ENST00000261448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214918.hg.1 |
NM_001232.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100214918.hg.1 |
ENST00000261448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214918.hg.1 |
NM_001232.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100214918.hg.1 |
TC0100015378.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100015378.hg.1 |
ENST00000261448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015378.hg.1 |
ENST00000261448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015378.hg.1 |
ENST00000261448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015378.hg.1 |
ENST00000261448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015378.hg.1 |
ENST00000261448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015378.hg.1 |
ENST00000261448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015378.hg.1 |
ENST00000261448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015378.hg.1 |
ENST00000261448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015378.hg.1 |
ENST00000261448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015378.hg.1 |
ENST00000261448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015378.hg.1 |
ENST00000261448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015378.hg.1 |
ENST00000261448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015378.hg.1 |
ENST00000261448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015378.hg.1 |
ENST00000261448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015378.hg.1 |
ENST00000261448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015378.hg.1 |
ENST00000261448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015378.hg.1 |
ENST00000488931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015378.hg.1 |
ENST00000488931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015378.hg.1 |
ENST00000488931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015378.hg.1 |
NM_001232.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015378.hg.1 |
NM_001232.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015378.hg.1 |
NM_001232.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015378.hg.1 |
NM_001232.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015378.hg.1 |
NM_001232.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015378.hg.1 |
NM_001232.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015378.hg.1 |
NM_001232.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015378.hg.1 |
NM_001232.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015378.hg.1 |
NM_001232.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015378.hg.1 |
NM_001232.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015378.hg.1 |
NM_001232.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015378.hg.1 |
NM_001232.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015378.hg.1 |
NM_001232.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015378.hg.1 |
NM_001232.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015378.hg.1 |
NM_001232.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015378.hg.1 |
NM_001232.3 |
Refseq |