Array Name |
Probe IDs |
Targets |
Target Type |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
38525 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
151614 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
970726 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
172275 |
AF106858.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
172275 |
AJ011001.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
172275 |
AK075270.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
172275 |
AK075555.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
172275 |
AK312405.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
172275 |
AY358400.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
172275 |
BC008770.2 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
172275 |
BC013207.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
172275 |
BT007311.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
172275 |
CR936747.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
172275 |
NM_005682.4 |
refseq_mrna_human_NM |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
172275 |
NM_201524.1 |
refseq_mrna_human_NM |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
172275 |
NM_201525.1 |
refseq_mrna_human_NM |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
109058 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
625463 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
820747 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
119589 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
420097 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
913503 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
271949 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
926545 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
426272 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
679743 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3233189 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_HS_44k_1947 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_HS_44k_1948 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
47238 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P02495486 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P02495498 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P19330826 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_18_P11192537 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_18_P11193886 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3233189 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_33_P3233189 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28379578 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07953860 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07953850 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07953837 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28379598 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07953843 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07953847 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28379600 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28379594 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28379577 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28379601 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28379596 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07953852 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28379588 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07953842 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07953845 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28379589 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28379590 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07953844 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28379591 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28379599 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07953846 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28379592 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07953843 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28379589 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07953842 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28379591 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28379601 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07953847 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07953845 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28379600 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28379577 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28379598 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07953846 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28379590 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07953837 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28379594 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28379599 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07953844 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28379592 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28379596 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28379578 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07953850 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07953852 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28379588 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P07953860 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953825 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16491379 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953702 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953727 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953827 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953683 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953715 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953839 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953704 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953691 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953763 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953794 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953713 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953851 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953760 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953782 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16491382 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953657 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953777 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953822 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16491390 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953806 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953775 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953861 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953709 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953749 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953765 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953761 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953699 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953755 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953793 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953797 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953694 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953724 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953848 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953751 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953809 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953820 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953843 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953790 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953845 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953772 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953787 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953729 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953744 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953796 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953675 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953785 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953853 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953811 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953661 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953665 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953711 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953668 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953779 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953819 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953676 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953678 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953768 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953817 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953721 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953846 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953733 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953659 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953707 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953753 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953770 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953739 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953813 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953836 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953857 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953685 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953747 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953682 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953736 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953780 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953815 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953718 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953834 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953671 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953679 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953802 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953742 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953746 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953800 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953842 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953758 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953689 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953737 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953696 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953741 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953832 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P07953858 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P19330888 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P02495498 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P39453097 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P19330832 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P02495508 |
- |
- |