Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057227.hg.1 |
TC01003728.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057227.hg.1 |
ENST00000308302.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057227.hg.1 |
NM_198447.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017022.hg.1 |
ENST00000308302.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017022.hg.1 |
ENST00000308302.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017022.hg.1 |
ENST00000308302.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017022.hg.1 |
ENST00000308302.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017022.hg.1 |
ENST00000308302.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017022.hg.1 |
ENST00000308302.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017022.hg.1 |
ENST00000308302.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017022.hg.1 |
ENST00000308302.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017022.hg.1 |
ENST00000308302.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017022.hg.1 |
ENST00000475517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017022.hg.1 |
ENST00000475517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017022.hg.1 |
ENST00000475517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017022.hg.1 |
NM_198447.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017022.hg.1 |
NM_198447.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017022.hg.1 |
NM_198447.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017022.hg.1 |
NM_198447.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017022.hg.1 |
NM_198447.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017022.hg.1 |
NM_198447.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017022.hg.1 |
NM_198447.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017022.hg.1 |
NM_198447.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017022.hg.1 |
NM_198447.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057228.hg.1 |
TC01003728.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057228.hg.1 |
ENST00000308302.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057228.hg.1 |
NM_198447.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057229.hg.1 |
TC01003728.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057229.hg.1 |
ENST00000308302.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057229.hg.1 |
NM_198447.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228954.hg.1 |
ENST00000308302.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228954.hg.1 |
NM_198447.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228954.hg.1 |
ENST00000308302.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228954.hg.1 |
NM_198447.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228954.hg.1 |
TC0100017022.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100228955.hg.1 |
ENST00000308302.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228955.hg.1 |
NM_198447.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228955.hg.1 |
ENST00000308302.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228955.hg.1 |
NM_198447.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228955.hg.1 |
TC0100017022.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100228956.hg.1 |
ENST00000308302.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228956.hg.1 |
NM_198447.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228956.hg.1 |
ENST00000308302.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228956.hg.1 |
NM_198447.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228956.hg.1 |
TC0100017022.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100017022.hg.1 |
ENST00000308302.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017022.hg.1 |
ENST00000308302.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017022.hg.1 |
ENST00000308302.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017022.hg.1 |
ENST00000308302.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017022.hg.1 |
ENST00000308302.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017022.hg.1 |
ENST00000308302.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017022.hg.1 |
ENST00000308302.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017022.hg.1 |
ENST00000308302.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017022.hg.1 |
ENST00000308302.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017022.hg.1 |
ENST00000475517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017022.hg.1 |
ENST00000475517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017022.hg.1 |
ENST00000475517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017022.hg.1 |
NM_198447.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017022.hg.1 |
NM_198447.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017022.hg.1 |
NM_198447.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017022.hg.1 |
NM_198447.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017022.hg.1 |
NM_198447.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017022.hg.1 |
NM_198447.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017022.hg.1 |
NM_198447.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017022.hg.1 |
NM_198447.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017022.hg.1 |
NM_198447.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228953.hg.1 |
ENST00000308302.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228953.hg.1 |
NM_198447.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228953.hg.1 |
ENST00000308302.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228953.hg.1 |
NM_198447.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228953.hg.1 |
TC0100017022.hg.1 |
Clariom D TC |