Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008449.hg.1 |
TC03000391.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008449.hg.1 |
ENST00000343837.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008449.hg.1 |
ENST00000496807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008449.hg.1 |
NM_001080537.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008450.hg.1 |
TC03000391.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008450.hg.1 |
ENST00000343837.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008450.hg.1 |
ENST00000496807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008450.hg.1 |
NM_001080537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155460.hg.1 |
ENST00000343837.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155460.hg.1 |
ENST00000469440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155460.hg.1 |
ENST00000496807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155460.hg.1 |
NM_001080537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155460.hg.1 |
ENST00000343837.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155460.hg.1 |
ENST00000469440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155460.hg.1 |
ENST00000496807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155460.hg.1 |
NM_001080537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155460.hg.1 |
TC0300007673.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300155461.hg.1 |
ENST00000343837.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155461.hg.1 |
ENST00000469440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155461.hg.1 |
ENST00000496807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155461.hg.1 |
NM_001080537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155461.hg.1 |
ENST00000343837.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155461.hg.1 |
ENST00000469440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155461.hg.1 |
ENST00000496807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155461.hg.1 |
NM_001080537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155461.hg.1 |
TC0300007673.hg.1 |
Clariom D TC |