Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004843.hg.1 |
ENST00000269053.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004843.hg.1 |
ENST00000394638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004843.hg.1 |
ENST00000485015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004843.hg.1 |
NM_173847.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004843.hg.1 |
TC17000381.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004845.hg.1 |
ENST00000269053.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004845.hg.1 |
ENST00000411740.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004845.hg.1 |
NM_173847.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004845.hg.1 |
TC17000381.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
ENST00000269053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
ENST00000269053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
ENST00000269053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
ENST00000269053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
ENST00000269053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
ENST00000269053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
ENST00000269053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
ENST00000269053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
ENST00000269053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
ENST00000394637.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
ENST00000394637.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
ENST00000394637.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
ENST00000394637.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
ENST00000394637.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
ENST00000394637.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
ENST00000394637.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
ENST00000394637.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
ENST00000394638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
ENST00000394638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
ENST00000394638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
ENST00000394638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
ENST00000394638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
ENST00000394638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
ENST00000394638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
ENST00000485015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
ENST00000485015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
ENST00000485015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
ENST00000580599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
ENST00000580599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
ENST00000580599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
ENST00000580599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
ENST00000580599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
ENST00000580599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
ENST00000580599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
ENST00000580599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
ENST00000580599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
NM_173847.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
NM_173847.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
NM_173847.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
NM_173847.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
NM_173847.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
NM_173847.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
NM_173847.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
NM_173847.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007536.hg.1 |
NM_173847.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004846.hg.1 |
ENST00000269053.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004846.hg.1 |
ENST00000411740.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004846.hg.1 |
ENST00000485015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004846.hg.1 |
NM_173847.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004846.hg.1 |
TC17000381.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700154258.hg.1 |
ENST00000269053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700154258.hg.1 |
ENST00000394638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700154258.hg.1 |
ENST00000485015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700154258.hg.1 |
NM_173847.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700154258.hg.1 |
ENST00000269053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700154258.hg.1 |
ENST00000394638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700154258.hg.1 |
ENST00000485015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700154258.hg.1 |
NM_173847.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700154258.hg.1 |
TC1700007536.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700154260.hg.1 |
ENST00000269053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700154260.hg.1 |
ENST00000394637.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700154260.hg.1 |
ENST00000580599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700154260.hg.1 |
NM_173847.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700154260.hg.1 |
ENST00000269053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700154260.hg.1 |
ENST00000394637.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700154260.hg.1 |
ENST00000580599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700154260.hg.1 |
NM_173847.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700154260.hg.1 |
TC1700007536.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700154261.hg.1 |
ENST00000269053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700154261.hg.1 |
ENST00000394637.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700154261.hg.1 |
ENST00000485015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700154261.hg.1 |
ENST00000580599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700154261.hg.1 |
NM_173847.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700154261.hg.1 |
ENST00000269053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700154261.hg.1 |
ENST00000394637.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700154261.hg.1 |
ENST00000485015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700154261.hg.1 |
ENST00000580599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700154261.hg.1 |
NM_173847.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700154261.hg.1 |
TC1700007536.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
ENST00000269053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
ENST00000269053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
ENST00000269053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
ENST00000269053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
ENST00000269053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
ENST00000269053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
ENST00000269053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
ENST00000269053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
ENST00000269053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
ENST00000394637.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
ENST00000394637.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
ENST00000394637.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
ENST00000394637.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
ENST00000394637.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
ENST00000394637.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
ENST00000394637.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
ENST00000394637.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
ENST00000394638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
ENST00000394638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
ENST00000394638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
ENST00000394638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
ENST00000394638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
ENST00000394638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
ENST00000394638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
ENST00000485015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
ENST00000485015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
ENST00000485015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
ENST00000580599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
ENST00000580599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
ENST00000580599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
ENST00000580599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
ENST00000580599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
ENST00000580599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
ENST00000580599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
ENST00000580599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
ENST00000580599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
NM_173847.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
NM_173847.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
NM_173847.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
NM_173847.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
NM_173847.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
NM_173847.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
NM_173847.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
NM_173847.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007536.hg.1 |
NM_173847.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700154257.hg.1 |
ENST00000269053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700154257.hg.1 |
ENST00000394638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700154257.hg.1 |
ENST00000485015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700154257.hg.1 |
NM_173847.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700154257.hg.1 |
ENST00000269053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700154257.hg.1 |
ENST00000394638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700154257.hg.1 |
ENST00000485015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700154257.hg.1 |
NM_173847.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700154257.hg.1 |
TC1700007536.hg.1 |
Clariom D TC |