Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010679.hg.1 |
ENST00000246662.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010679.hg.1 |
ENST00000246662.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010679.hg.1 |
ENST00000246662.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010679.hg.1 |
ENST00000246662.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010679.hg.1 |
ENST00000246662.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010679.hg.1 |
ENST00000246662.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010679.hg.1 |
ENST00000246662.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010679.hg.1 |
ENST00000246662.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010679.hg.1 |
ENST00000246662.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010679.hg.1 |
ENST00000246662.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010679.hg.1 |
ENST00000246662.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010679.hg.1 |
ENST00000246662.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010679.hg.1 |
ENST00000588431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010679.hg.1 |
ENST00000588431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010679.hg.1 |
ENST00000588431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010679.hg.1 |
ENST00000588431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010679.hg.1 |
ENST00000588431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010679.hg.1 |
ENST00000588431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010679.hg.1 |
ENST00000588431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010679.hg.1 |
ENST00000588431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010679.hg.1 |
ENST00000588431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010679.hg.1 |
ENST00000588431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010679.hg.1 |
NM_000226.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010679.hg.1 |
NM_000226.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010679.hg.1 |
NM_000226.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010679.hg.1 |
NM_000226.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010679.hg.1 |
NM_000226.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010679.hg.1 |
NM_000226.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010679.hg.1 |
NM_000226.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010679.hg.1 |
NM_000226.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010679.hg.1 |
NM_000226.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010679.hg.1 |
NM_000226.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010679.hg.1 |
NM_000226.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010679.hg.1 |
NM_000226.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17020021.hg.1 |
ENST00000246662.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17020021.hg.1 |
NM_000226.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17020021.hg.1 |
TC17001512.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700184476.hg.1 |
ENST00000246662.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184476.hg.1 |
ENST00000588431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184476.hg.1 |
NM_000226.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700184476.hg.1 |
ENST00000246662.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184476.hg.1 |
ENST00000588431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184476.hg.1 |
NM_000226.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700184476.hg.1 |
TC1700010679.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700010679.hg.1 |
ENST00000246662.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010679.hg.1 |
ENST00000246662.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010679.hg.1 |
ENST00000246662.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010679.hg.1 |
ENST00000246662.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010679.hg.1 |
ENST00000246662.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010679.hg.1 |
ENST00000246662.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010679.hg.1 |
ENST00000246662.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010679.hg.1 |
ENST00000246662.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010679.hg.1 |
ENST00000246662.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010679.hg.1 |
ENST00000246662.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010679.hg.1 |
ENST00000246662.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010679.hg.1 |
ENST00000246662.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010679.hg.1 |
ENST00000588431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010679.hg.1 |
ENST00000588431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010679.hg.1 |
ENST00000588431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010679.hg.1 |
ENST00000588431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010679.hg.1 |
ENST00000588431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010679.hg.1 |
ENST00000588431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010679.hg.1 |
ENST00000588431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010679.hg.1 |
ENST00000588431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010679.hg.1 |
ENST00000588431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010679.hg.1 |
ENST00000588431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010679.hg.1 |
NM_000226.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010679.hg.1 |
NM_000226.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010679.hg.1 |
NM_000226.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010679.hg.1 |
NM_000226.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010679.hg.1 |
NM_000226.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010679.hg.1 |
NM_000226.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010679.hg.1 |
NM_000226.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010679.hg.1 |
NM_000226.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010679.hg.1 |
NM_000226.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010679.hg.1 |
NM_000226.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010679.hg.1 |
NM_000226.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010679.hg.1 |
NM_000226.3 |
Refseq |