Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01048946.hg.1 |
TC01003195.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01048946.hg.1 |
ENST00000369145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01048946.hg.1 |
ENST00000369146.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01048946.hg.1 |
NM_014849.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
ENST00000369145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
ENST00000369145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
ENST00000369145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
ENST00000369145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
ENST00000369145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
ENST00000369145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
ENST00000369145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
ENST00000369145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
ENST00000369145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
ENST00000369145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
ENST00000369145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
ENST00000369145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
ENST00000369145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
ENST00000369145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
ENST00000369146.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
ENST00000369146.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
ENST00000369146.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
ENST00000369146.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
ENST00000369146.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
ENST00000369146.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
ENST00000369146.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
ENST00000369146.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
ENST00000369146.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
ENST00000369146.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
ENST00000369146.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
ENST00000369146.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
ENST00000369146.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
ENST00000369146.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
ENST00000369146.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
ENST00000369146.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
ENST00000369146.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
NM_001278719.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
NM_001278719.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
NM_001278719.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
NM_001278719.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
NM_001278719.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
NM_001278719.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
NM_014849.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
NM_014849.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
NM_014849.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
NM_014849.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
NM_014849.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
NM_014849.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
NM_014849.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
NM_014849.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
NM_014849.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
NM_014849.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
NM_014849.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
NM_014849.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
NM_014849.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
NM_014849.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
NM_014849.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
NM_014849.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015714.hg.1 |
NM_014849.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100217635.hg.1 |
ENST00000369145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100217635.hg.1 |
ENST00000369146.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100217635.hg.1 |
NM_014849.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100217635.hg.1 |
ENST00000369145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100217635.hg.1 |
ENST00000369146.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100217635.hg.1 |
NM_014849.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100217635.hg.1 |
TC0100015714.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
ENST00000369145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
ENST00000369145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
ENST00000369145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
ENST00000369145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
ENST00000369145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
ENST00000369145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
ENST00000369145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
ENST00000369145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
ENST00000369145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
ENST00000369145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
ENST00000369145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
ENST00000369145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
ENST00000369145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
ENST00000369145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
ENST00000369146.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
ENST00000369146.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
ENST00000369146.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
ENST00000369146.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
ENST00000369146.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
ENST00000369146.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
ENST00000369146.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
ENST00000369146.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
ENST00000369146.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
ENST00000369146.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
ENST00000369146.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
ENST00000369146.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
ENST00000369146.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
ENST00000369146.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
ENST00000369146.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
ENST00000369146.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
ENST00000369146.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
NM_001278719.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
NM_001278719.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
NM_001278719.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
NM_001278719.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
NM_001278719.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
NM_001278719.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
NM_014849.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
NM_014849.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
NM_014849.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
NM_014849.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
NM_014849.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
NM_014849.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
NM_014849.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
NM_014849.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
NM_014849.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
NM_014849.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
NM_014849.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
NM_014849.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
NM_014849.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
NM_014849.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
NM_014849.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
NM_014849.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015714.hg.1 |
NM_014849.4 |
Refseq |