Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018760.hg.1 |
TC01001211.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018760.hg.1 |
ENST00000271636.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018760.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018763.hg.1 |
TC01001211.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018763.hg.1 |
ENST00000271636.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018763.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000416743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000416743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000416743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000416743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000427934.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000427934.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000427934.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000427934.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000427934.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000464886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000464886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000464886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000464886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000467998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000467998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000467998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000467998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000467998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000473377.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000473377.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000473377.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000473377.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000473377.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000473377.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000473377.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000473377.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000473377.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000473377.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000502442.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000502442.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000505188.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
ENST00000505188.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172162.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172162.hg.1 |
ENST00000427934.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172162.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172162.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172162.hg.1 |
ENST00000427934.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172162.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172162.hg.1 |
TC0100009958.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100172165.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172165.hg.1 |
ENST00000427934.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172165.hg.1 |
ENST00000502442.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172165.hg.1 |
ENST00000505188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172165.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172165.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172165.hg.1 |
ENST00000427934.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172165.hg.1 |
ENST00000502442.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172165.hg.1 |
ENST00000505188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172165.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172165.hg.1 |
TC0100009958.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000416743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000416743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000416743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000416743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000427934.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000427934.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000427934.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000427934.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000427934.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000464886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000464886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000464886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000464886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000467998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000467998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000467998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000467998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000467998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000473377.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000473377.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000473377.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000473377.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000473377.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000473377.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000473377.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000473377.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000473377.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000473377.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000502442.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000502442.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000505188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
ENST00000505188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009958.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172161.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172161.hg.1 |
ENST00000427934.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172161.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172161.hg.1 |
ENST00000271636.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172161.hg.1 |
ENST00000427934.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172161.hg.1 |
NM_020770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172161.hg.1 |
TC0100009958.hg.1 |
Clariom D TC |