Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007757.hg.1 |
ENST00000603021.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004490.hg.1 |
TC04000319.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004490.hg.1 |
ENST00000264228.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004490.hg.1 |
NM_024592.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004491.hg.1 |
TC04000319.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004491.hg.1 |
ENST00000264228.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004491.hg.1 |
NM_024592.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007556.hg.1 |
ENST00000264228.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007556.hg.1 |
ENST00000264228.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007556.hg.1 |
ENST00000264228.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007556.hg.1 |
ENST00000264228.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007556.hg.1 |
ENST00000264228.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007556.hg.1 |
ENST00000264228.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007556.hg.1 |
ENST00000264228.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007556.hg.1 |
ENST00000264228.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007556.hg.1 |
ENST00000264228.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007556.hg.1 |
ENST00000264228.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007556.hg.1 |
ENST00000264228.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007556.hg.1 |
ENST00000505210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007556.hg.1 |
ENST00000505210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007556.hg.1 |
ENST00000505210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007556.hg.1 |
ENST00000505210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007556.hg.1 |
ENST00000505210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007556.hg.1 |
ENST00000514398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007556.hg.1 |
ENST00000514398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007556.hg.1 |
ENST00000514398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007556.hg.1 |
ENST00000514398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007556.hg.1 |
NM_024592.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007556.hg.1 |
NM_024592.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007556.hg.1 |
NM_024592.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007556.hg.1 |
NM_024592.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007556.hg.1 |
NM_024592.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007556.hg.1 |
NM_024592.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007556.hg.1 |
NM_024592.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007556.hg.1 |
NM_024592.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007556.hg.1 |
NM_024592.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007556.hg.1 |
NM_024592.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004494.hg.1 |
TC04000319.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004494.hg.1 |
ENST00000264228.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004494.hg.1 |
ENST00000514398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004494.hg.1 |
NM_024592.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004493.hg.1 |
TC04000319.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004493.hg.1 |
ENST00000264228.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004493.hg.1 |
ENST00000514398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004493.hg.1 |
NM_024592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400151660.hg.1 |
ENST00000264228.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151660.hg.1 |
ENST00000505210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151660.hg.1 |
ENST00000514398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151660.hg.1 |
NM_024592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400151660.hg.1 |
ENST00000264228.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151660.hg.1 |
ENST00000505210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151660.hg.1 |
ENST00000514398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151660.hg.1 |
NM_024592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400151660.hg.1 |
TC0400007556.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100007757.hg.1 |
ENST00000603021.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151661.hg.1 |
ENST00000264228.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151661.hg.1 |
ENST00000514398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151661.hg.1 |
NM_024592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400151661.hg.1 |
ENST00000264228.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151661.hg.1 |
ENST00000514398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151661.hg.1 |
NM_024592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400151661.hg.1 |
TC0400007556.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400151656.hg.1 |
ENST00000264228.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151656.hg.1 |
NM_024592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400151656.hg.1 |
ENST00000264228.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151656.hg.1 |
NM_024592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400151656.hg.1 |
TC0400007556.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400151657.hg.1 |
ENST00000264228.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151657.hg.1 |
NM_024592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400151657.hg.1 |
ENST00000264228.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151657.hg.1 |
NM_024592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400151657.hg.1 |
TC0400007556.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400007556.hg.1 |
ENST00000264228.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007556.hg.1 |
ENST00000264228.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007556.hg.1 |
ENST00000264228.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007556.hg.1 |
ENST00000264228.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007556.hg.1 |
ENST00000264228.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007556.hg.1 |
ENST00000264228.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007556.hg.1 |
ENST00000264228.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007556.hg.1 |
ENST00000264228.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007556.hg.1 |
ENST00000264228.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007556.hg.1 |
ENST00000264228.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007556.hg.1 |
ENST00000264228.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007556.hg.1 |
ENST00000505210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007556.hg.1 |
ENST00000505210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007556.hg.1 |
ENST00000505210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007556.hg.1 |
ENST00000505210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007556.hg.1 |
ENST00000505210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007556.hg.1 |
ENST00000514398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007556.hg.1 |
ENST00000514398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007556.hg.1 |
ENST00000514398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007556.hg.1 |
ENST00000514398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007556.hg.1 |
NM_024592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007556.hg.1 |
NM_024592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007556.hg.1 |
NM_024592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007556.hg.1 |
NM_024592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007556.hg.1 |
NM_024592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007556.hg.1 |
NM_024592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007556.hg.1 |
NM_024592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007556.hg.1 |
NM_024592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007556.hg.1 |
NM_024592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007556.hg.1 |
NM_024592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400151658.hg.1 |
ENST00000264228.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151658.hg.1 |
ENST00000505210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151658.hg.1 |
NM_024592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400151658.hg.1 |
ENST00000264228.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151658.hg.1 |
ENST00000505210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151658.hg.1 |
NM_024592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400151658.hg.1 |
TC0400007556.hg.1 |
Clariom D TC |