Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08182324 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612699 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612717 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612713 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612710 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612711 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612753 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612666 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612703 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612683 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612722 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612738 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08182278 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612725 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612681 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08182327 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08182315 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612734 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612689 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612674 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612667 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612726 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612670 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612694 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08182273 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612707 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612737 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612756 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08182252 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612704 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612682 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612701 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612754 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612700 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08182316 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612668 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612669 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612748 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612706 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612672 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612715 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612678 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16529944 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612744 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612730 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612724 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16529941 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612695 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612728 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612665 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612747 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612727 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08182305 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08182298 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612718 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612739 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28612671 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08182277 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
111103 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
4016 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 8x60K |
38916 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 8x60K |
15662 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
554689 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
141143 |
AK314397.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
141143 |
BC001273.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
141143 |
BC050685.2 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
141143 |
BC056880.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
141143 |
CR592211.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
141143 |
CR592671.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
141143 |
CR596991.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
141143 |
CR601378.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
141143 |
CR602422.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
141143 |
CR611350.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
141143 |
CR616462.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
141143 |
CR616589.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
141143 |
DQ233668.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
141143 |
L41668.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
141143 |
NM_000403.3 |
refseq_mrna_human_NM |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
141143 |
NM_001008216.1 |
refseq_mrna_human_NM |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
329512 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3258977 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3352578 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_44k_23_P25235 |
- |
- |
Agilent Whole Human Genome Oligo Microarray 4x44K |
19146 |
- |
- |
Agilent Human Genome CGH Microarray 44B |
A_14_P103906 |
- |
- |
Agilent Human Genome CGH Microarray 44B |
A_14_P200773 |
- |
- |
Agilent Human Genome CGH Microarray 44A |
A_14_P103906 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
32812 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
48149 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P33119644 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P39577861 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P02566971 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P39577881 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_14_P103906 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P19451301 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3258977 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3352578 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_33_P3352578 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_33_P3258977 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612674 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612694 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612685 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612706 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612710 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612713 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612703 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612675 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16529941 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612676 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612701 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612679 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612699 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08182278 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612708 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612683 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612678 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612689 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16529946 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612717 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612682 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612704 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612695 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08182266 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08182272 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612707 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612715 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16529944 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612717 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612689 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08182266 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16529941 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612695 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612713 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612703 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612704 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612706 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08182272 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612694 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612715 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08182278 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612682 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612685 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612674 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612708 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612710 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612701 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612683 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16529944 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16529946 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612678 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612707 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612675 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612679 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612699 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28612676 |
- |
- |
Whole Human Genome Oligo Microarray G4112A |
A_23_P25235 |
- |
- |
Agilent Human Genome CGH Microarray 105A (G4412A) |
A_14_P200773 |
- |
- |
Agilent Human Genome CGH Microarray 105A (G4412A) |
A_14_P103906 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08182277 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08182288 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08182266 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08182296 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08182275 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08182278 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08182290 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08182271 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08182282 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08182294 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08182286 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08182300 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08182284 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08182292 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08182270 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08182298 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08182272 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08182269 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08182264 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08182274 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_14_P200773 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_14_P103906 |
- |
- |
Agilent Human Genome CGH Microarray 4x44K |
A_14_P200773 |
- |
- |
Agilent Human Genome CGH Microarray 4x44K |
A_14_P103906 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K G4112F |
A_23_P25235 |
- |
- |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
91602 |
- |
- |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
19715 |
- |
- |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
47483 |
- |
- |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
85009 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_14_P103906 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P39577881 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_14_P200773 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182298 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182249 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182284 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182332 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182261 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182340 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182251 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182301 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182274 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182272 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182314 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182316 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182347 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182269 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182256 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182264 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182286 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182294 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182292 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182277 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182309 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182296 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182337 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182344 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182330 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182335 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182318 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182300 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182271 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182306 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182327 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182262 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182325 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182288 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182282 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182303 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182320 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182254 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182259 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182312 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182322 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182290 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08182266 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_14_P200773 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_14_P103906 |
- |
- |