Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011532.hg.1 |
TC08000867.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011532.hg.1 |
ENST00000331434.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011532.hg.1 |
NM_023080.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011534.hg.1 |
TC08000867.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011534.hg.1 |
ENST00000331434.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011534.hg.1 |
NM_023080.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000331434.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000331434.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000331434.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000331434.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000331434.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000331434.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000331434.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000331434.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000331434.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000331434.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000524395.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000524395.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000524395.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000524395.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000524395.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000524395.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000524395.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000524395.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000524395.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000529593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000529593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000529593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000529593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000530455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000530455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000530455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000530455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000530455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000530455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000530455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000530455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000530455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000534350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000534350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000534350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000534350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000534350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000534350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
ENST00000534350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
NM_023080.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
NM_023080.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
NM_023080.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
NM_023080.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
NM_023080.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
NM_023080.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
NM_023080.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
NM_023080.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009301.hg.1 |
NM_023080.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800163821.hg.1 |
ENST00000331434.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163821.hg.1 |
ENST00000524395.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163821.hg.1 |
ENST00000529593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163821.hg.1 |
ENST00000530455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163821.hg.1 |
NM_023080.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800163821.hg.1 |
ENST00000331434.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163821.hg.1 |
ENST00000524395.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163821.hg.1 |
ENST00000529593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163821.hg.1 |
ENST00000530455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800163821.hg.1 |
NM_023080.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800163821.hg.1 |
TC0800009301.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000331434.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000331434.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000331434.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000331434.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000331434.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000331434.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000331434.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000331434.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000331434.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000331434.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000524395.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000524395.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000524395.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000524395.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000524395.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000524395.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000524395.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000524395.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000524395.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000529593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000529593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000529593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000529593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000530455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000530455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000530455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000530455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000530455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000530455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000530455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000530455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000530455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000534350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000534350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000534350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000534350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000534350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000534350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
ENST00000534350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
NM_023080.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
NM_023080.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
NM_023080.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
NM_023080.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
NM_023080.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
NM_023080.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
NM_023080.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
NM_023080.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009301.hg.1 |
NM_023080.2 |
Refseq |