Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019341.hg.1 |
NM_152219.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019341.hg.1 |
TC17001461.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019345.hg.1 |
NM_152219.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019345.hg.1 |
TC17001461.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007793.hg.1 |
ENST00000578774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007793.hg.1 |
ENST00000578774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007793.hg.1 |
ENST00000578774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010615.hg.1 |
ENST00000578689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010615.hg.1 |
ENST00000578689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010615.hg.1 |
NM_152219.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010615.hg.1 |
NM_152219.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010615.hg.1 |
NM_152219.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010615.hg.1 |
NM_152219.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010615.hg.1 |
NM_152219.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010615.hg.1 |
NM_152219.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019343.hg.1 |
ENST00000337376.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019343.hg.1 |
NM_152219.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019343.hg.1 |
TC17001461.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700183686.hg.1 |
NM_152219.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700183686.hg.1 |
NM_152219.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700183686.hg.1 |
TC1700010615.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700007793.hg.1 |
ENST00000578774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007793.hg.1 |
ENST00000578774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007793.hg.1 |
ENST00000578774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700183683.hg.1 |
ENST00000578689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700183683.hg.1 |
NM_152219.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700183683.hg.1 |
ENST00000578689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700183683.hg.1 |
NM_152219.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700183683.hg.1 |
TC1700010615.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700010615.hg.1 |
ENST00000578689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010615.hg.1 |
ENST00000578689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010615.hg.1 |
NM_152219.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010615.hg.1 |
NM_152219.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010615.hg.1 |
NM_152219.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010615.hg.1 |
NM_152219.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010615.hg.1 |
NM_152219.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010615.hg.1 |
NM_152219.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700183684.hg.1 |
ENST00000578689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700183684.hg.1 |
NM_152219.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700183684.hg.1 |
ENST00000578689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700183684.hg.1 |
NM_152219.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700183684.hg.1 |
TC1700010615.hg.1 |
Clariom D TC |