Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014195.hg.1 |
ENST00000375790.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014195.hg.1 |
ENST00000419761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014195.hg.1 |
NM_001242702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014195.hg.1 |
NM_173576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014195.hg.1 |
TC10001133.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014197.hg.1 |
ENST00000375790.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014197.hg.1 |
ENST00000419761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014197.hg.1 |
NM_001242702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014197.hg.1 |
NM_173576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014197.hg.1 |
TC10001133.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
ENST00000375790.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
ENST00000375790.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
ENST00000375790.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
ENST00000375790.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
ENST00000375790.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
ENST00000375790.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
ENST00000375790.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
ENST00000375790.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
ENST00000375790.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
ENST00000375790.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
ENST00000375790.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
ENST00000419761.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
ENST00000419761.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
ENST00000419761.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
ENST00000419761.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
ENST00000419761.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
ENST00000419761.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
ENST00000419761.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
ENST00000419761.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
ENST00000419761.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
ENST00000419761.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
ENST00000460919.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
ENST00000460919.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
ENST00000460919.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
ENST00000460919.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
ENST00000460919.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
ENST00000460919.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
ENST00000460919.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
ENST00000561227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
ENST00000561227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
ENST00000561227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
NM_001242702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
NM_001242702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
NM_001242702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
NM_001242702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
NM_001242702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
NM_001242702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
NM_001242702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
NM_001242702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
NM_001242702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
NM_001242702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
NM_173576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
NM_173576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
NM_173576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
NM_173576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
NM_173576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
NM_173576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
NM_173576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
NM_173576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
NM_173576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010132.hg.1 |
NM_173576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014199.hg.1 |
ENST00000375790.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014199.hg.1 |
ENST00000419761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014199.hg.1 |
NM_001242702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014199.hg.1 |
NM_173576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014199.hg.1 |
TC10001133.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000169055.hg.1 |
ENST00000375790.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169055.hg.1 |
ENST00000419761.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169055.hg.1 |
ENST00000460919.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169055.hg.1 |
ENST00000561227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169055.hg.1 |
NM_001242702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000169055.hg.1 |
NM_173576.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000169055.hg.1 |
ENST00000375790.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169055.hg.1 |
ENST00000419761.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169055.hg.1 |
ENST00000460919.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169055.hg.1 |
ENST00000561227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169055.hg.1 |
NM_001242702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000169055.hg.1 |
NM_173576.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000169055.hg.1 |
TC1000010132.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000169059.hg.1 |
ENST00000375790.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169059.hg.1 |
ENST00000419761.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169059.hg.1 |
NM_001242702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000169059.hg.1 |
NM_173576.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000169059.hg.1 |
ENST00000375790.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169059.hg.1 |
ENST00000419761.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169059.hg.1 |
NM_001242702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000169059.hg.1 |
NM_173576.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000169059.hg.1 |
TC1000010132.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000169054.hg.1 |
ENST00000375790.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169054.hg.1 |
ENST00000419761.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169054.hg.1 |
ENST00000460919.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169054.hg.1 |
ENST00000561227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169054.hg.1 |
NM_001242702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000169054.hg.1 |
NM_173576.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000169054.hg.1 |
ENST00000375790.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169054.hg.1 |
ENST00000419761.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169054.hg.1 |
ENST00000460919.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169054.hg.1 |
ENST00000561227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169054.hg.1 |
NM_001242702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000169054.hg.1 |
NM_173576.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000169054.hg.1 |
TC1000010132.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000169060.hg.1 |
ENST00000375790.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169060.hg.1 |
ENST00000419761.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169060.hg.1 |
NM_001242702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000169060.hg.1 |
NM_173576.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000169060.hg.1 |
ENST00000375790.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169060.hg.1 |
ENST00000419761.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169060.hg.1 |
NM_001242702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000169060.hg.1 |
NM_173576.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000169060.hg.1 |
TC1000010132.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
ENST00000375790.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
ENST00000375790.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
ENST00000375790.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
ENST00000375790.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
ENST00000375790.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
ENST00000375790.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
ENST00000375790.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
ENST00000375790.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
ENST00000375790.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
ENST00000375790.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
ENST00000375790.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
ENST00000419761.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
ENST00000419761.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
ENST00000419761.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
ENST00000419761.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
ENST00000419761.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
ENST00000419761.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
ENST00000419761.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
ENST00000419761.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
ENST00000419761.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
ENST00000419761.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
ENST00000460919.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
ENST00000460919.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
ENST00000460919.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
ENST00000460919.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
ENST00000460919.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
ENST00000460919.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
ENST00000460919.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
ENST00000561227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
ENST00000561227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
ENST00000561227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
NM_001242702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
NM_001242702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
NM_001242702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
NM_001242702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
NM_001242702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
NM_001242702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
NM_001242702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
NM_001242702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
NM_001242702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
NM_001242702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
NM_173576.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
NM_173576.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
NM_173576.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
NM_173576.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
NM_173576.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
NM_173576.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
NM_173576.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
NM_173576.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
NM_173576.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010132.hg.1 |
NM_173576.2 |
Refseq |