Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22013308.hg.1 |
ENST00000303592.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22013308.hg.1 |
NM_152868.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22013308.hg.1 |
TC22000790.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008701.hg.1 |
ENST00000303592.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008701.hg.1 |
ENST00000303592.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008701.hg.1 |
ENST00000303592.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008701.hg.1 |
ENST00000303592.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008701.hg.1 |
ENST00000303592.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008701.hg.1 |
NM_004981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008701.hg.1 |
NM_004981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008701.hg.1 |
NM_004981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008701.hg.1 |
NM_004981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008701.hg.1 |
NM_004981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008701.hg.1 |
NM_152868.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008701.hg.1 |
NM_152868.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008701.hg.1 |
NM_152868.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008701.hg.1 |
NM_152868.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008701.hg.1 |
NM_152868.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22013305.hg.1 |
ENST00000303592.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22013305.hg.1 |
NM_004981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22013305.hg.1 |
NM_152868.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22013305.hg.1 |
TC22000790.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22013306.hg.1 |
ENST00000303592.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22013306.hg.1 |
NM_004981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22013306.hg.1 |
NM_152868.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22013306.hg.1 |
TC22000790.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2200162982.hg.1 |
TC2200008701.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200162982.hg.1 |
ENST00000303592.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200162982.hg.1 |
NM_004981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200162982.hg.1 |
NM_152868.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200162982.hg.1 |
ENST00000303592.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200162982.hg.1 |
NM_004981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200162982.hg.1 |
NM_152868.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200162985.hg.1 |
TC2200008701.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200162985.hg.1 |
ENST00000303592.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200162985.hg.1 |
NM_152868.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200162985.hg.1 |
ENST00000303592.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200162985.hg.1 |
NM_152868.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200162983.hg.1 |
TC2200008701.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200162983.hg.1 |
ENST00000303592.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200162983.hg.1 |
NM_004981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200162983.hg.1 |
NM_152868.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200162983.hg.1 |
ENST00000303592.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200162983.hg.1 |
NM_004981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200162983.hg.1 |
NM_152868.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008701.hg.1 |
ENST00000303592.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008701.hg.1 |
ENST00000303592.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008701.hg.1 |
ENST00000303592.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008701.hg.1 |
ENST00000303592.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008701.hg.1 |
ENST00000303592.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008701.hg.1 |
NM_004981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008701.hg.1 |
NM_004981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008701.hg.1 |
NM_004981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008701.hg.1 |
NM_004981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008701.hg.1 |
NM_004981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008701.hg.1 |
NM_152868.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008701.hg.1 |
NM_152868.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008701.hg.1 |
NM_152868.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008701.hg.1 |
NM_152868.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008701.hg.1 |
NM_152868.2 |
Refseq |