Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
ENST00000349552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
ENST00000349552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
ENST00000349552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
ENST00000349552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
ENST00000349552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
ENST00000349552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
ENST00000349552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
ENST00000349552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
ENST00000349552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
ENST00000349552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
ENST00000349552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
ENST00000349552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
ENST00000349552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
ENST00000349552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
ENST00000349552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
ENST00000542375.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
ENST00000542375.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
ENST00000542375.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
ENST00000542375.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
ENST00000542375.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
ENST00000542375.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
ENST00000542375.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
ENST00000542375.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
ENST00000542375.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
ENST00000542375.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
ENST00000542375.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
ENST00000542375.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
ENST00000542375.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
ENST00000542375.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
ENST00000542375.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
ENST00000542375.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
NM_174897.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
NM_174897.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
NM_174897.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
NM_174897.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
NM_174897.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
NM_174897.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
NM_174897.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
NM_174897.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
NM_174897.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
NM_174897.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
NM_174897.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
NM_174897.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
NM_174897.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
NM_174897.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007139.hg.1 |
NM_174897.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002752.hg.1 |
ENST00000349552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002752.hg.1 |
NM_174897.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002752.hg.1 |
TC20000221.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000148875.hg.1 |
ENST00000349552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148875.hg.1 |
ENST00000542375.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148875.hg.1 |
NM_174897.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148875.hg.1 |
TC2000007139.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000148875.hg.1 |
ENST00000349552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148875.hg.1 |
ENST00000542375.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148875.hg.1 |
NM_174897.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
ENST00000349552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
ENST00000349552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
ENST00000349552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
ENST00000349552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
ENST00000349552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
ENST00000349552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
ENST00000349552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
ENST00000349552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
ENST00000349552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
ENST00000349552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
ENST00000349552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
ENST00000349552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
ENST00000349552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
ENST00000349552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
ENST00000349552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
ENST00000542375.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
ENST00000542375.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
ENST00000542375.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
ENST00000542375.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
ENST00000542375.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
ENST00000542375.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
ENST00000542375.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
ENST00000542375.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
ENST00000542375.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
ENST00000542375.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
ENST00000542375.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
ENST00000542375.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
ENST00000542375.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
ENST00000542375.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
ENST00000542375.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
ENST00000542375.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
NM_174897.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
NM_174897.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
NM_174897.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
NM_174897.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
NM_174897.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
NM_174897.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
NM_174897.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
NM_174897.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
NM_174897.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
NM_174897.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
NM_174897.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
NM_174897.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
NM_174897.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
NM_174897.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007139.hg.1 |
NM_174897.2 |
Refseq |