Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001415.hg.1 |
TC01000090.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001415.hg.1 |
ENST00000328191.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001415.hg.1 |
ENST00000333172.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001415.hg.1 |
ENST00000351136.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001415.hg.1 |
ENST00000437392.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001415.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001415.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001416.hg.1 |
TC01000090.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001416.hg.1 |
ENST00000328191.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001416.hg.1 |
ENST00000333172.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001416.hg.1 |
ENST00000351136.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001416.hg.1 |
ENST00000437392.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001416.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001416.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
ENST00000328191.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
ENST00000328191.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
ENST00000328191.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
ENST00000328191.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
ENST00000351136.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
ENST00000351136.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
ENST00000351136.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
ENST00000351136.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
ENST00000351136.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
ENST00000351136.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
ENST00000351136.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
ENST00000351136.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
ENST00000351136.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
ENST00000351136.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
ENST00000411823.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
ENST00000411823.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
ENST00000411823.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
ENST00000411823.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
ENST00000415267.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
ENST00000415267.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
ENST00000415267.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
ENST00000415267.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006695.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001414.hg.1 |
TC01000090.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001414.hg.1 |
ENST00000328191.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001414.hg.1 |
ENST00000333172.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001414.hg.1 |
ENST00000351136.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001414.hg.1 |
ENST00000437392.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001414.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001414.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001413.hg.1 |
TC01000090.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001413.hg.1 |
ENST00000328191.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001413.hg.1 |
ENST00000333172.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001413.hg.1 |
ENST00000437392.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001413.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001413.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100146337.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100146337.hg.1 |
ENST00000351136.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100146337.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100146337.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100146337.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100146337.hg.1 |
ENST00000351136.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100146337.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100146337.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100146337.hg.1 |
TC0100006695.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100146335.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100146335.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100146335.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100146335.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100146335.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100146335.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100146335.hg.1 |
TC0100006695.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
ENST00000328191.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
ENST00000328191.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
ENST00000328191.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
ENST00000328191.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
ENST00000351136.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
ENST00000351136.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
ENST00000351136.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
ENST00000351136.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
ENST00000351136.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
ENST00000351136.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
ENST00000351136.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
ENST00000351136.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
ENST00000351136.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
ENST00000351136.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
ENST00000411823.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
ENST00000411823.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
ENST00000411823.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
ENST00000411823.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
ENST00000415267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
ENST00000415267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
ENST00000415267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
ENST00000415267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006695.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100146339.hg.1 |
ENST00000328191.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100146339.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100146339.hg.1 |
ENST00000351136.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100146339.hg.1 |
ENST00000411823.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100146339.hg.1 |
ENST00000415267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100146339.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100146339.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100146339.hg.1 |
ENST00000328191.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100146339.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100146339.hg.1 |
ENST00000351136.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100146339.hg.1 |
ENST00000411823.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100146339.hg.1 |
ENST00000415267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100146339.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100146339.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100146339.hg.1 |
TC0100006695.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100146336.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100146336.hg.1 |
ENST00000351136.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100146336.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100146336.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100146336.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100146336.hg.1 |
ENST00000351136.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100146336.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100146336.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100146336.hg.1 |
TC0100006695.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100146338.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100146338.hg.1 |
ENST00000351136.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100146338.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100146338.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100146338.hg.1 |
ENST00000333172.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100146338.hg.1 |
ENST00000351136.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100146338.hg.1 |
NM_138697.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100146338.hg.1 |
NM_177540.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100146338.hg.1 |
TC0100006695.hg.1 |
Clariom D TC |