Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003239.hg.1 |
ENST00000354760.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003239.hg.1 |
NM_001886.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003239.hg.1 |
TC22000171.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006955.hg.1 |
ENST00000354760.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006955.hg.1 |
ENST00000354760.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006955.hg.1 |
ENST00000354760.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006955.hg.1 |
ENST00000354760.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006955.hg.1 |
ENST00000354760.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006955.hg.1 |
ENST00000354760.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006955.hg.1 |
ENST00000354760.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006955.hg.1 |
ENST00000354760.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006955.hg.1 |
ENST00000354760.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006955.hg.1 |
ENST00000466315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006955.hg.1 |
ENST00000466315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006955.hg.1 |
ENST00000466315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006955.hg.1 |
ENST00000466315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006955.hg.1 |
ENST00000466315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006955.hg.1 |
ENST00000466315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006955.hg.1 |
ENST00000466315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006955.hg.1 |
NM_001886.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006955.hg.1 |
NM_001886.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006955.hg.1 |
NM_001886.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006955.hg.1 |
NM_001886.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006955.hg.1 |
NM_001886.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006955.hg.1 |
NM_001886.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006955.hg.1 |
NM_001886.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006955.hg.1 |
NM_001886.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006955.hg.1 |
NM_001886.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003238.hg.1 |
ENST00000354760.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003238.hg.1 |
ENST00000466315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003238.hg.1 |
NM_001886.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003238.hg.1 |
TC22000171.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2200148357.hg.1 |
TC2200006955.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200148357.hg.1 |
ENST00000354760.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200148357.hg.1 |
NM_001886.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200148357.hg.1 |
ENST00000354760.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200148357.hg.1 |
NM_001886.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200148356.hg.1 |
TC2200006955.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200148356.hg.1 |
ENST00000354760.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200148356.hg.1 |
ENST00000466315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200148356.hg.1 |
NM_001886.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200148356.hg.1 |
ENST00000354760.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200148356.hg.1 |
ENST00000466315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200148356.hg.1 |
NM_001886.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006955.hg.1 |
ENST00000354760.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006955.hg.1 |
ENST00000354760.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006955.hg.1 |
ENST00000354760.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006955.hg.1 |
ENST00000354760.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006955.hg.1 |
ENST00000354760.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006955.hg.1 |
ENST00000354760.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006955.hg.1 |
ENST00000354760.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006955.hg.1 |
ENST00000354760.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006955.hg.1 |
ENST00000354760.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006955.hg.1 |
ENST00000466315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006955.hg.1 |
ENST00000466315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006955.hg.1 |
ENST00000466315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006955.hg.1 |
ENST00000466315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006955.hg.1 |
ENST00000466315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006955.hg.1 |
ENST00000466315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006955.hg.1 |
ENST00000466315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006955.hg.1 |
NM_001886.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006955.hg.1 |
NM_001886.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006955.hg.1 |
NM_001886.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006955.hg.1 |
NM_001886.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006955.hg.1 |
NM_001886.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006955.hg.1 |
NM_001886.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006955.hg.1 |
NM_001886.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006955.hg.1 |
NM_001886.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006955.hg.1 |
NM_001886.2 |
Refseq |