Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016933.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016933.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016933.hg.1 |
TC12001297.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010108.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200176360.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176360.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200176360.hg.1 |
ENST00000261195.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176360.hg.1 |
NM_021957.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200176360.hg.1 |
TC1200010108.hg.1 |
Clariom D TC |