Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008951.hg.1 |
ENST00000216951.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008951.hg.1 |
ENST00000216951.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008951.hg.1 |
ENST00000216951.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008951.hg.1 |
ENST00000216951.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008951.hg.1 |
ENST00000216951.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008951.hg.1 |
ENST00000216951.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008951.hg.1 |
ENST00000216951.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008951.hg.1 |
ENST00000216951.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008951.hg.1 |
ENST00000216951.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008951.hg.1 |
ENST00000216951.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008951.hg.1 |
ENST00000216951.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008951.hg.1 |
ENST00000216951.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008951.hg.1 |
ENST00000216951.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008951.hg.1 |
ENST00000216951.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008951.hg.1 |
ENST00000451957.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008951.hg.1 |
ENST00000451957.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008951.hg.1 |
ENST00000451957.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008951.hg.1 |
ENST00000451957.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008951.hg.1 |
ENST00000451957.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008951.hg.1 |
ENST00000451957.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008951.hg.1 |
ENST00000451957.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008951.hg.1 |
ENST00000451957.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008951.hg.1 |
ENST00000451957.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008951.hg.1 |
NM_000178.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008951.hg.1 |
NM_000178.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008951.hg.1 |
NM_000178.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008951.hg.1 |
NM_000178.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008951.hg.1 |
NM_000178.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008951.hg.1 |
NM_000178.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008951.hg.1 |
NM_000178.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008951.hg.1 |
NM_000178.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008951.hg.1 |
NM_000178.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008951.hg.1 |
NM_000178.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008951.hg.1 |
NM_000178.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008951.hg.1 |
NM_000178.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008951.hg.1 |
NM_000178.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008951.hg.1 |
NM_000178.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010546.hg.1 |
ENST00000216951.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010546.hg.1 |
ENST00000451957.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010546.hg.1 |
ENST00000541098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010546.hg.1 |
NM_000178.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010546.hg.1 |
TC20000789.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010545.hg.1 |
ENST00000216951.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010545.hg.1 |
ENST00000451957.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010545.hg.1 |
ENST00000541098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010545.hg.1 |
NM_000178.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010545.hg.1 |
TC20000789.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000162327.hg.1 |
ENST00000216951.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000162327.hg.1 |
ENST00000451957.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000162327.hg.1 |
NM_000178.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000162327.hg.1 |
TC2000008951.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000162327.hg.1 |
ENST00000216951.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000162327.hg.1 |
ENST00000451957.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000162327.hg.1 |
NM_000178.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008951.hg.1 |
ENST00000216951.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008951.hg.1 |
ENST00000216951.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008951.hg.1 |
ENST00000216951.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008951.hg.1 |
ENST00000216951.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008951.hg.1 |
ENST00000216951.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008951.hg.1 |
ENST00000216951.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008951.hg.1 |
ENST00000216951.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008951.hg.1 |
ENST00000216951.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008951.hg.1 |
ENST00000216951.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008951.hg.1 |
ENST00000216951.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008951.hg.1 |
ENST00000216951.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008951.hg.1 |
ENST00000216951.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008951.hg.1 |
ENST00000216951.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008951.hg.1 |
ENST00000216951.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008951.hg.1 |
ENST00000451957.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008951.hg.1 |
ENST00000451957.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008951.hg.1 |
ENST00000451957.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008951.hg.1 |
ENST00000451957.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008951.hg.1 |
ENST00000451957.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008951.hg.1 |
ENST00000451957.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008951.hg.1 |
ENST00000451957.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008951.hg.1 |
ENST00000451957.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008951.hg.1 |
ENST00000451957.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008951.hg.1 |
NM_000178.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008951.hg.1 |
NM_000178.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008951.hg.1 |
NM_000178.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008951.hg.1 |
NM_000178.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008951.hg.1 |
NM_000178.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008951.hg.1 |
NM_000178.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008951.hg.1 |
NM_000178.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008951.hg.1 |
NM_000178.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008951.hg.1 |
NM_000178.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008951.hg.1 |
NM_000178.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008951.hg.1 |
NM_000178.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008951.hg.1 |
NM_000178.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008951.hg.1 |
NM_000178.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008951.hg.1 |
NM_000178.3 |
Refseq |