Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15009884.hg.1 |
ENST00000353809.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15009884.hg.1 |
ENST00000354638.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15009884.hg.1 |
ENST00000382996.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15009884.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15009884.hg.1 |
TC15001091.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000431101.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000431101.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000431101.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000431101.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000431101.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000431101.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000431101.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000431101.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000445578.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000445578.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000445578.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000445578.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000445578.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000445578.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
ENST00000445578.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500163502.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500163502.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500163502.hg.1 |
ENST00000445578.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500163502.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500163502.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500163502.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500163502.hg.1 |
ENST00000445578.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500163502.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500163502.hg.1 |
TC1500008794.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000353809.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000354638.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000431101.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000431101.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000431101.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000431101.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000431101.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000431101.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000431101.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000431101.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000445578.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000445578.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000445578.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000445578.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000445578.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000445578.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
ENST00000445578.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008794.hg.1 |
NM_000275.2 |
Refseq |