Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACVR2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106.3 | NP_001097.2 | 9 - 10 | 1236 | 61 | |
XM_005265583.3 | XP_005265640.1 | 9 - 10 | 3698 | 61 | ||
XM_017007514.1 | XP_016863003.1 | 9 - 10 | 3677 | 61 | ||
XM_017007515.1 | XP_016863004.1 | 9 - 10 | 1322 | 61 | ||
XM_017007516.1 | XP_016863005.1 | 9 - 10 | 4646 | 61 | ||
GenBank mRNA | AB209449.1 | - | 8 - 9 | 5001 | 61 | |
BC096243.1 | - | 9 - 10 | 1216 | 40005760 | 61 | |
BC096244.1 | - | 9 - 10 | 1216 | 40005761 | 61 | |
BC096245.1 | - | 8 - 9 | 1298 | 40005763 | 61 | |
BC099642.4 | - | 9 - 10 | 1216 | 40005762 | 61 | |
X77533.1 | - | 9 - 10 | 1216 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |