Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318789.1 | NP_001305718.1 | 2 - 3 | 250 | 80 | |
NM_001318790.1 | NP_001305719.1 | 2 - 3 | 227 | 80 | ||
NM_001318791.1 | NP_001305720.1 | 2 - 3 | 216 | 80 | ||
NM_001318793.1 | NP_001305722.1 | 2 - 3 | 212 | 80 | ||
NM_003264.4 | NP_003255.2 | 2 - 3 | 235 | 80 | ||
GenBank mRNA | AF051152.1 | - | 1 - 2 | 141 | 80 | |
BI769109.1 | - | 2 - 3 | 198 | 80 | ||
DA460298.1 | - | 3 - 4 | 201 | 80 | ||
U88878.1 | - | 1 - 2 | 108 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Inventoried Probe spans exons |