Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022906.hg.1 |
ENST00000247829.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022906.hg.1 |
ENST00000393330.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022906.hg.1 |
ENST00000546561.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022906.hg.1 |
ENST00000552128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022906.hg.1 |
NM_004616.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022906.hg.1 |
TC12001723.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000247829.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000247829.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000247829.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000247829.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000247829.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000247829.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000247829.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000247829.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000247829.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000247829.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000247829.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000247829.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000247829.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000247829.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000247829.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000393330.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000393330.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000393330.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000393330.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000393330.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000393330.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000393330.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000393330.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000393330.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000393330.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000393330.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000393330.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000393330.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000393330.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000393330.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000393330.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000546561.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000546561.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000546561.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000546561.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000546561.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000546561.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000546561.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000546561.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000546561.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000546561.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000546561.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000546561.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000546561.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000549421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000549421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000549421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000549421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000550818.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000550818.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000550818.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000550818.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000550818.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000550818.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000552128.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000552128.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000552128.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000552128.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000552128.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000552128.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000552128.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000552786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000552786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000552786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000552786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000552786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
ENST00000552786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
NM_004616.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
NM_004616.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
NM_004616.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
NM_004616.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
NM_004616.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
NM_004616.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
NM_004616.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
NM_004616.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
NM_004616.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
NM_004616.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
NM_004616.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
NM_004616.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
NM_004616.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011251.hg.1 |
NM_004616.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000247829.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000247829.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000247829.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000247829.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000247829.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000247829.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000247829.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000247829.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000247829.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000247829.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000247829.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000247829.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000247829.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000247829.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000247829.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000393330.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000393330.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000393330.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000393330.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000393330.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000393330.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000393330.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000393330.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000393330.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000393330.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000393330.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000393330.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000393330.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000393330.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000393330.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000393330.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000546561.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000546561.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000546561.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000546561.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000546561.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000546561.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000546561.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000546561.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000546561.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000546561.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000546561.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000546561.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000546561.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000549421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000549421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000549421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000549421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000550818.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000550818.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000550818.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000550818.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000550818.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000550818.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000552128.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000552128.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000552128.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000552128.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000552128.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000552128.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000552128.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000552786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000552786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000552786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000552786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000552786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
ENST00000552786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
NM_004616.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
NM_004616.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
NM_004616.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
NM_004616.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
NM_004616.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
NM_004616.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
NM_004616.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
NM_004616.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
NM_004616.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
NM_004616.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
NM_004616.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
NM_004616.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
NM_004616.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011251.hg.1 |
NM_004616.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200187227.hg.1 |
ENST00000247829.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200187227.hg.1 |
ENST00000393330.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200187227.hg.1 |
ENST00000546561.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200187227.hg.1 |
ENST00000552128.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200187227.hg.1 |
NM_004616.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200187227.hg.1 |
ENST00000247829.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200187227.hg.1 |
ENST00000393330.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200187227.hg.1 |
ENST00000546561.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200187227.hg.1 |
ENST00000552128.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200187227.hg.1 |
NM_004616.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200187227.hg.1 |
TC1200011251.hg.1 |
Clariom D TC |