Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024201.hg.1 |
TC17002855.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024201.hg.1 |
ENST00000307078.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024201.hg.1 |
ENST00000375702.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024201.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000375702.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000375702.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000375702.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000375702.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000375702.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000375702.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000375702.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000375702.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000375702.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000375702.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000375702.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000375702.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000375702.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000375702.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000375702.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000375702.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000544103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000544103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000544103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000544103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000544103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000577278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000577278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000577278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000577278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000577278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000577278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000577278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000578251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000578251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000578251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000578251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000580513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000580513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000580513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000580513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000580513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000580513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000585045.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000585045.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000585045.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000585045.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000585045.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000585045.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000611991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000611991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000611991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000611991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000611991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000611991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000307078.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000375702.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000375702.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000375702.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000375702.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000375702.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000375702.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000375702.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000375702.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000375702.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000375702.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000375702.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000375702.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000375702.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000375702.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000375702.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000375702.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000544103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000544103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000544103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000544103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000544103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000577278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000577278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000577278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000577278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000577278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000577278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000577278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000578251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000578251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000578251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000578251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000580513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000580513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000580513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000580513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000580513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000580513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000585045.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000585045.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000585045.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000585045.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000585045.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000585045.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000611991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000611991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000611991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000611991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000611991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000611991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
ENST00000618960.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012457.hg.1 |
NM_004655.3 |
Refseq |